Skip to Content
Merck
All Photos(1)

Key Documents

33942

Supelco

Quizalofop-P-tefuryl solution

100 μg/mL in acetonitrile, PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H21ClN2O5
CAS Number:
Molecular Weight:
428.87
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

concentration

100 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

single component solution

storage temp.

2-8°C

SMILES string

C[C@@H](Oc1ccc(Oc2cnc3cc(Cl)ccc3n2)cc1)C(=O)OCC4CCCO4

InChI

1S/C22H21ClN2O5/c1-14(22(26)28-13-18-3-2-10-27-18)29-16-5-7-17(8-6-16)30-21-12-24-20-11-15(23)4-9-19(20)25-21/h4-9,11-12,14,18H,2-3,10,13H2,1H3/t14-,18?/m1/s1

InChI key

BBKDWPHJZANJGB-IKJXHCRLSA-N

General description

Quizalofop-P-tefuryl is a selective, post-emergence herbicide used for the control of annual and perennial grasses in potato, sugar beet, sunflower, oilseed rape, peanut and vegetable crops. Its mode of action involves the inhibition of acetyl CoA carboxylase activity.

Application

Quizalofop-P-tefuryl has been used as an analytical reference standard for the quantification of the analyte in soil samples using ultra high- performance liquid chromatography coupled to orbitrap mass spectrometry (UHPLC-Orbitrap-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Degradation studies of quizalofop-p and related compounds in soils using liquid chromatography coupled to low and high resolution mass analyzers.
Lopez-Ruiz R, et al.
The Science of the Total Environment, 607(1), 204-213 (2017)
Rosalía López-Ruiz et al.
Ecotoxicology and environmental safety, 157, 285-291 (2018-04-09)
A degradation study of quizalofop-p and its commercial products (quizalofop-p-ethyl, quizalofop-p-tefuryl and propaquizafop) in water samples has been performed using ultra high-performance liquid chromatography coupled to Orbitrap mass spectrometry (UHPLC-Orbitrap-MS). CHHQ (dihydroxychloroquinoxalin), CHQ (6-chloroquinoxalin-2-ol) and PPA ((R)-2-(4-hydroxyphenoxy)propionicacid) were the main
Fate and persistence of herbicide quizalofop-p-tefuryl on black gram.
Mukhopadhya S, et al.
Journal of Crop and Weed, 8(1), 190-192 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service