Skip to Content
Merck
All Photos(1)

Key Documents

33553-M

Sigma-Aldrich

Pyridine

puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N
CAS Number:
Molecular Weight:
79.10
Beilstein:
103233
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

reag. Ph. Eur.

vapor density

2.72 (vs air)

vapor pressure

10 mmHg ( 13.2 °C)
20 mmHg ( 25 °C)

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

899 °F

expl. lim.

12.4 %

impurities

≤0.0005% KMnO4 red. matter (as O)
≤0.002% ammonia (NH3)
≤0.002% non-volatile matter
≤0.01% piperidine (GC)
≤0.1% water (Karl Fischer)
≤0.2% 2-picoline (GC)

refractive index

n20/D 1.509 (lit.)

pH

8.5 (25 °C, 15.82 g/L)

bp

115 °C (lit.)

mp

−42 °C (lit.)

solubility

H2O: in accordance

density

0.978 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤5 mg/kg
sulfate (SO42-): ≤5 mg/kg

cation traces

Al: ≤0.5 mg/kg
B: ≤0.02 mg/kg
Ba: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.03 mg/kg
Fe: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sn: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

C1=CN=CC=C1

InChI

1S/C5H5N/c1-2-4-6-5-3-1/h1-5H

InChI key

JUJWROOIHBZHMG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyridine is a basic N-heterocyclic compound. It acts as a nitrogen donor ligand and forms many metal-pyridine complexes. Its complexes having tetrahedral and octahedral geometries can be differentiated by infrared spectral investigations.

Application

Pyridine (py) may be used in the preparation of vanadium oxide nanoparticles and iron benzenedicarboxylates, Fe(OH)(BDC)(py)0.85 (BDC = 1,4-benzenedicarboxylate, py = pyridine). It may be employed as a catalyst in Knoevenagel condensation reaction.

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

62.6 °F - closed cup

Flash Point(C)

17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service