Skip to Content
Merck
All Photos(2)

Key Documents

33197

Supelco

Haloxyfop-P-methyl

PESTANAL®, analytical standard

Synonym(s):

(R)-2-{4-[3-Chloro-5-(trifluoromethyl)-2-pyridyloxy]phenoxy}propanoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13ClF3NO4
CAS Number:
Molecular Weight:
375.73
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COC(=O)[C@@H](C)Oc1ccc(Oc2ncc(cc2Cl)C(F)(F)F)cc1

InChI

1S/C16H13ClF3NO4/c1-9(15(22)23-2)24-11-3-5-12(6-4-11)25-14-13(17)7-10(8-21-14)16(18,19)20/h3-9H,1-2H3/t9-/m1/s1

InChI key

MFSWTRQUCLNFOM-SECBINFHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Haloxyfop-P-methyl is a selective postemergence aryloxyphenoxypropionate (AOPP) herbicide.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effectiveness of Clodinafop-Propargyl, Haloxyfop-p-methyl and Difenzoquat-methyl-sulfate Plus [TM="Adigor"] and [TM="Propel"] Adjuvants in Controlling Avena ludoviciana Durieu.
Parsa M, et al.
Journal of Agriculture, Science and Technology, 16(2), 291-299 (2014)
Hepatotoxicity, nephrotoxicity and oxidative stress in rat testis following exposure to haloxyfop-p-methyl ester, an aryloxyphenoxypropionate herbicide.
Olayinka TB and Ore A
Toxics, 3(4), 373-389 (2015)
Sunwoo Park et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 233, 108761-108761 (2020-04-15)
Haloxyfop-P-methyl, an aryloxyphenoxypropionate herbicide, is widely used to eliminate unwanted plants by inhibiting lipid synthesis and inducing oxidative stress. Since haloxyfop-P-methyl targets are limited within plants, few negative side effects on non-target crops have been reported. However, dissolved haloxyfop-P-methyl in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service