Skip to Content
Merck
All Photos(2)

Documents

230251

Sigma-Aldrich

Eosin Y

75% (HPLC)

Synonym(s):

2′,4′,5′,7′-Tetrabromofluorescein, Acid Red 87, Bromo acid J. TS, XL, or XX, Bromofluorescein, Bronze Bromo ES, Eosin yellowish, Solvent red 43

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H8Br4O5
CAS Number:
Molecular Weight:
647.89
Colour Index Number:
45380:2
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

75% (HPLC)

form

powder

color

orange to very dark orange

solubility

spirit: soluble

λmax

524 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1c(Br)cc2c(Oc3c(Br)c(O)c(Br)cc3C24OC(=O)c5ccccc45)c1Br

InChI

1S/C20H8Br4O5/c21-11-5-9-17(13(23)15(11)25)28-18-10(6-12(22)16(26)14(18)24)20(9)8-4-2-1-3-7(8)19(27)29-20/h1-6,25-26H

InChI key

DBZJJPROPLPMSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Eosin Y is a xanthene dye and is used for the differential staining of connective tissue and cytoplasm. In histopathology, it is applied as a counterstain after hematoxylin and before methylene blue. It is also used as a background stain, thereby giving contrast to the nuclear stains.

Application

Eosin Y has been used as a reagent in the preparation of Nigrosin-Eosin (NE) stain and is used in hematoxylin and eosin (H&E) staining technique to visualize alginate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gregorios J B
Histophatologic Techniques (2006)
Morphometric dimensions of the spermatozoa in Thai native boar depend onthe ejaculates
Nasinprom N
Animal Reproduction, 540-543 (2015)
Concordance among pathologists in the second Cardiac Allograft Rejection Gene Expression Observational Study (CARGO II).
Crespo-Leiro MG, Zuckermann A, Bara C, et al.
Transplantation, 94(11), 1172-1177 (2012)
Electrophoretic detection of alginate by Hematoxylin and Eosin fluorescence: Implications in cell encapsulation/tissue engineering
Verma Y K
Reactive functional Polymers, 102, 130-136 (2016)
John D. Bancroft
Theory and Practice of Histological Techniques (2008)

Protocols

The article describes the sample preparation and photometric determination of silver concentration in water samples using Spectroquant® photometers and test kits.

The article describes the sample preparation and photometric determination of silver concentration in water samples using Spectroquant® photometers and test kits.

The article describes the sample preparation and photometric determination of silver concentration in water samples using Spectroquant® photometers and test kits.

The article describes the sample preparation and photometric determination of silver concentration in water samples using Spectroquant® photometers and test kits.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service