Skip to Content
Merck
All Photos(1)

Key Documents

11363

Sigma-Aldrich

Avicel® PH-101

tested according to Ph. Eur.

Synonym(s):

Cellulose, Cellulose microcrystalline, Cellulosum microcristallinum, Cellulose powder, Cotton linters

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.21

Agency

USP/NF
tested according to Ph. Eur.

Quality Level

form

powder

color

white

useful pH range

5.0 - 7.5

solubility

soluble (ammoniacal solution of copper tetramine)

SMILES string

O1[C@H](C(C(C(C1CO)O)O)O)O[C@@H]2C(OC(C(C2O)O)O)CO

InChI

1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3?,4?,5?,6?,7?,8?,9?,10-,11?,12+/m1/s1

InChI key

GUBGYTABKSRVRQ-WFVLMXAXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

High purity cellulose powders for partition chromatography.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Legal Information

Avicel is a registered trademark of FMC Corp.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Hu et al.
Wei sheng wu xue bao = Acta microbiologica Sinica, 53(4), 354-362 (2013-07-19)
Use of renewable plant biomass is an active area in current biotechnology research. This report explores the cellulose-degradation system and the factors affecting cellulase gene expression in Chaetomium globosum NK102. In the sequenced genome, we identified 10 cellulase genes by
Jung Min Lee et al.
JAMA otolaryngology-- head & neck surgery, 139(6), 629-635 (2013-06-22)
Bioresorbable fixation systems have been popular for the treatment of facial fractures. However, their mechanical properties are uncertain and complications have been reported. To overcome these problems, we developed a bioresorbable fixation plate using a composite of silk fibroin and
Battle for the barrel.
Robert F Service
Science (New York, N.Y.), 339(6126), 1374-1377 (2013-03-23)
Orly Salama-Alber et al.
The Journal of biological chemistry, 288(23), 16827-16838 (2013-04-13)
The rumen bacterium Ruminococcus flavefaciens produces a highly organized multienzyme cellulosome complex that plays a key role in the degradation of plant cell wall polysaccharides, notably cellulose. The R. flavefaciens cellulosomal system is anchored to the bacterial cell wall through
Jerome M Fox et al.
Nature chemical biology, 9(6), 356-361 (2013-04-09)
The mechanisms of enzyme activity on solid substrates are not well understood. Unlike enzyme catalysis in aqueous solutions, enzyme activity on surfaces is complicated by adsorption steps and structural heterogeneities that make enzyme-substrate interactions difficult to characterize. Cellulase enzymes, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service