Skip to Content
Merck
All Photos(1)

Key Documents

MABS1910

Sigma-Aldrich

Anti-Signal peptide peptidase-like 3 Antibody, clone 7F9

clone 7F9, from mouse

Synonym(s):

Signal peptide peptidase-like 3, SPP-like 3, Intramembrane protease 2, IMP-2, Presenilin homologous protein 1, PSH1, Presenilin-like protein 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
7F9, monoclonal
application:
IP
WB
species reactivity:
human
technique(s):
immunoprecipitation (IP): suitable
western blot: suitable
citations:
2

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7F9, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... SPPL3(121665)

General description

Signal peptide peptidase-like 3 (UniProt: Q8TCT6; also known as SPP-like 3, Intramembrane protease 2, IMP-2, Presenilin homologous protein 1, PSH1, Presenilin-like protein 4, SPPL3) is encoded by the SPPL3 (also known as IMP2) gene (Gene ID: 121665) in human. SPPL3 is a multi-pass transmembrane protein that is shown to be widely expressed and is highly conserved in multicellular eukaryotes. Three isoforms of SPPL3 have been described that are produced by alternative splicing. It is a member of the intramembrane-cleaving GxGD proteases. It acts as an intramembrane protease that cleaves type II membrane protein substrates in or close to their luminal transmembrane domain boundaries. It can also act as a sheddase by mediating the proteolytic release and secretion of active site-containing ectodomains of glycan-modifying glycosidases and glycosyltransferases. It is also shown to catalyze the intramembrane cleavage of the envelope glycoprotein gp130 and/or the leader peptide gp18LP of the simian foamy virus independent of prior ectodomain shedding by furin or furin-like proprotein convertase-mediated cleavage proteolysis. The first transmembrane domain of SPPL3 may act as a type I signal anchor. SPPL3 also contains a PAL motif (aa 342-344) that is required for normal active site conformation.

Specificity

Clone 7F9 detects Signal peptide peptidase-like 3 in human cells. it targets an epitope within 15 amino acids from the C-terminal half. Immunogen sequence is conserved in all three isoforms.

Immunogen

KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal half of human Signal peptide peptidase-like 3.

Application

Anti-Signal peptide peptidase-like 3, clone 7F9, Cat. No. MABS1910, is a mouse monoclonal antibody that detects Signal peptide peptidase-like 3 and has been tested for use in Immunoprecipitation and Western Blotting.
Research Category
Signaling
Western Blotting Analysis: A representative lot detected Signal peptide peptidase-like 3 in Western Blotting applications (Voss, M., et. al. (2012). J Biol Chem. 287(52):43401-9; Voss, M., et. al. (2014). EMBO J. 33(24):2890-905; Fleck, D., et. al. (2016). J Biol Chem. 291(1):318-33).

Immunoprecipitation Analysis: A representative lot detected Signal peptide peptidase-like 3 in Immunoprecipitation applications (Voss, M., et. al. (2012). J Biol Chem. 287(52):43401-9).

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Signal peptide peptidase-like 3 in HEK293 cell lysate.

Target description

~42 kDa observed; 42.56 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amanda Heard et al.
Nature communications, 13(1), 3367-3367 (2022-06-12)
While chimeric antigen receptor (CAR) T cells targeting CD19 can cure a subset of patients with B cell malignancies, most patients treated will not achieve durable remission. Identification of the mechanisms leading to failure is essential to broadening the efficacy
Xiaoxuan Zhuang et al.
Cell reports, 43(4), 114105-114105 (2024-04-15)
Natural killer (NK) cells are primary defenders against cancer precursors, but cancer cells can persist by evading immune surveillance. To investigate the genetic mechanisms underlying this evasion, we perform a genome-wide CRISPR screen using B lymphoblastoid cells. SPPL3, a peptidase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service