Skip to Content
Merck
All Photos(1)

Key Documents

MABS1812M

Sigma-Aldrich

Anti-Furin Antibody, clone 16B1.1

clone 16B1.1, from mouse

Synonym(s):

Anti-FUR, Anti-PACE, Anti-PCSK3, Anti-SPC1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

16B1.1, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... FURIN(5045)

General description

Furin (UniProt: P09958; also known as EC: 3.4.2.75, Dibasic-processing enzyme, Paired basic amino acid residue-cleaving enzyme, PACE) is encoded by the FURIN (also known as FUR, PACE, PCSK3) gene (Gene ID: 5045) in human. Furin is an ubiquitously expressed enzyme with endoprotease activity within constitutive secretory pathways and is capable of cleavage at the RX(K/R)R consensus motif. It is single-pass type I membrane protein that shuttles between the trans-Golgi network and the cell surface. It is synthesized with a signal peptide of 26 amino acids and a propeptide region (aa 27-107) that inhibits its activity. The inhibition peptide plays the role of an intramolecular chaperone. It is autocatalytically removed in the endoplasmic reticulum (ER) and remains non-covalently bound to furin as a potent autoinhibitor. Propeptide cleavage is a prerequisite for exit of Furin molecules out of the endoplasmic reticulum. A second cleavage within the propeptide region occurs in the trans-Golgi network, followed by the release of the propeptide and the activation of Furin. Mature Furin has a luminal region (aa 108-705), a helical domain (aa 716-738), and a cytoplasmic region (aa 739-794). The cytoplasmic domain responsible for its trans-Golgi network localization and recycling from the cell surface.

Specificity

Clone 16B1.1 detects human Furin and it targets an epitope within 177 amino acids from the C-terminal half.

Immunogen

GST/His-tagged recombinant fragment corresponding to 177 amino acids from the C-terminal half of human Furin.

Application

Anti-Furin, clone 16B1.1, Cat. No. MABS1812, is a mouse monoclonal antibody that detects Furin in human cells and has been tested for use in Western Blotting.

Quality

Evaluated by Western Blotting in U-251 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Furin in 10 µg of U-251 cell lysate.

Target description

~100 kDa observed; 86.68 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service