Skip to Content
Merck
All Photos(1)

Key Documents

MABN2470

Sigma-Aldrich

Anti-Aquaporin-4 Antibody

mouse monoclonal, mCCD

Synonym(s):

AQP-4, Mercurial-insensitive water channel, MIWC, WCH4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Aquaporin-4 Antibody, clone mCCD, clone mCCD, from mouse

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

mCCD, monoclonal

species reactivity

human, rat, mouse

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG3κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... AQP4(361)

General description

Aquaporin-4 (UniProt: P55087; also known as AQP-4, Mercurial-insensitive water channel, MIWC, WCH4) is encoded by the AGP4 gene (Gene ID: 361) in human. Aquaporin-4 is a multi-pass membrane protein that forms a water-specific channel and serves as an osmoreceptor that regulates body water balance and mediates water flow within the central nervous system. Its extracellular domains are localized to amino acids 58-64, 137-155, and 206-231); its transmembrane domains are in amino acids 37-57, 65-85, 116-136, 156-176, 185-205, and 232-252); and the cytoplasmic domain is in amino acids 1-36, 86-115, 177-184, and 253-323). Aquaporin-4 is largely expressed in the brain and the skeletal muscle and at lower levels in heart, kidney, and lung tissue. Phosphorylation of Aquaporin-4 at Serine 180 by protein kinase C is shown to reduce conductance by about 50%, whereas phosphorylation at Serine 111 by protein kinase G in response to glutamate can increase conductance by about 40%. Two isoforms of Aquaporin-4 have been described that are produced by alternative splicing. Antibodies to Aquaporin-4 can serve as useful serum markers to distinguishing neuromyelitis optica (NM) from multiple sclerosis. Clone mCCD does not cause redistribution of Aquaporin-4 and also it does not affect coaggregation astrocytic CD32-B and Aquaporin-4. (Ref.: Hinson, SR et al. (2017). Proc. Natl. Acad. Sci. USA 114(21); 5491-5496).

Specificity

Clone mCCD specifically detects Aquaporin-4 in human, mouse, and rat brain tissue sections. It targets an epitope with in 71 amino acids from the cytoplasmic domain from the C-terminal region.

Immunogen

Epitope: cytoplasmic domain
His-tagged full length human recombinant Aquaporin-4.

Application

Anti-Aquaporin-4, clone mCCD Antibody, Cat. No. MABN2470, is a highly specific mouse monoclonal antibody that targets Aquaporin-4 and has been tested for use in Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, and Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis: A representative lot detected Aquaporin-4 in Western Blotting applications (Hinson, S.R., et. al. (2017). Proc Natl Acad Sci U S A. 114(21):5491-5496).

Immunoprecipitation Analysis: A representative lot immunoprecipitated Aquaporin-4 in Immunoprecipitation applications (Hinson, S.R., et. al. (2017). Proc Natl Acad Sci U S A. 114(21):5491-5496).

Immunocytochemistry Analysis: A representative lot detected Aquaporin-4 in Immunocytochemistry applications (Hinson, S.R., et. al. (2017). Proc Natl Acad Sci U S A. 114(21):5491-5496).

Quality

Evaluated by Immunohistochemistry in rat brain frozen tissue sections.

Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected Aquaporin-4 in rat brain frozen tissue sections.

Target description

34.83 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG3 in PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service