Skip to Content
Merck
All Photos(1)

Key Documents

MABD24A4

Sigma-Aldrich

Anti-NANOG Antibody, clone 7F7.1, Alexa Fluor 488 conjugate

clone 7F7.1, from mouse, ALEXA FLUOR 488

Synonym(s):

Homeobox protein NANOG, Homeobox transcription factor Nanog, hNanog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7F7.1, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NANOG(79923)

General description

NANOG (Homeobox protein NANOG) is a member of the Nanog homeobox family of DNA-binding proteins. It is expressed in embryonic stem cells and confers pluripotency on these cells. Once embryonic stem cells become differentiated, NANOG expression is suppressed. NANOG is involved in the Hedgehog/Gli1 signaling pathway which has been implicated in the development and growth of various types of tumors. NANOG has also been identified as a key transcription factor used to generate induced pluripotent stem cells.

Application

Anti-NANOG Antibody, clone 7F7.1, Alexa Fluor 488 conjugate.

Quality

Evaluated by Immunocytochemistry in H9 human embryonic stem cells. Immunocytochemsitry Analysis: A 1:100 dilution of this antibody detected NANOG in H9 human embryonic stem cells.

Target description

The uncojugated parent antibody (Catalog No. MABD24) has an observed MW of 39 kDa

Physical form

Purified mouse monoclonal IgG2a conjugated to Alexa Fluor 488 in PBS with 0.1% sodium azide and 15mg/ml BSA.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Breanna S Borys et al.
Stem cells translational medicine, 9(9), 1036-1052 (2020-05-24)
Human induced pluripotent stem cells (hiPSCs) have generated a great deal of attention owing to their capacity for self-renewal and differentiation into the three germ layers of the body. Their discovery has facilitated a new era in biomedicine for understanding
Breanna S Borys et al.
Stem cell research & therapy, 12(1), 55-55 (2021-01-14)
Human induced pluripotent stem cells (hiPSCs) hold enormous promise in accelerating breakthroughs in understanding human development, drug screening, disease modeling, and cell and gene therapies. Their potential, however, has been bottlenecked in a mostly laboratory setting due to bioprocess challenges
Victor J T Lin et al.
Scientific reports, 7(1), 5005-5005 (2017-07-12)
Despite their well-known function in maintaining normal cell physiology, how inorganic elements are relevant to cellular pluripotency and differentiation in human pluripotent stem cells (hPSCs) has yet to be systematically explored. Using total reflection X-ray fluorescence (TXRF) spectrometry and inductively
Quan Qi et al.
International journal of molecular medicine, 35(3), 569-578 (2014-12-20)
The present study aimed to investigate the X chromosome inactivation (XCI) status in long-term cultured human parthenogenetic embryonic stem cells. One human embryonic stem (hES) cell line and 2 human parthenogenetic embryonic stem (hPES) cell lines were subjected to long-term
Melissa Conti Mazza et al.
Stem cell research, 55, 102506-102506 (2021-08-23)
Mutations in the oncogene PARK7, which codes for DJ-1, have been associated with early-onset autosomal recessive Parkinson's disease (PD); however, the exact role of DJ-1 in PD remains elusive. Fibroblasts from a PD patient with a uniparental disomy, 1 bp deletion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service