Skip to Content
Merck
All Photos(1)

Key Documents

MAB80138

Sigma-Aldrich

Anti-Human Metapneumovirus Antibody, clone 507

clone 507, Chemicon®, from mouse

Synonym(s):

hMPV

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

507, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG2a

shipped in

wet ice

Related Categories

General description

Human metapneumovirus (hMPV) is a member of the pneumovirinae subfamily of paramyxoviruses, first described in 2001 from pediatric respiratory specimens in the Netherlands. It is an enveloped pleomorphic virus with a single negative-strand RNA genome. Two major serotypes, A and B, have been described. Several studies identified hMPV in specimens worldwide, and estimated that by age 5 years 70% of children have developed antibodies to hMPV.

Specificity

Recognizes the nuclear protein. Reactive with all hMPV sub-types; A1, A2, B1, B2.

Application

Detect Human Metapneumovirus using this Anti-Human Metapneumovirus Antibody, clone 507 validated for use in ELISA, FC, IF & WB.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoyan Li et al.
Intervirology, 58(6), 403-412 (2016-04-21)
No licensed vaccines or therapeutic agents for human metapneumovirus (hMPV) infection exist to date. We aimed to construct a multi-epitope peptide (MEP) of hMPV to show promising results for epitope-based vaccine development. Six independent algorithms were screened to predict B-cell
Simon Loevenich et al.
Mediators of inflammation, 2019, 4964239-4964239 (2019-11-07)
Human metapneumovirus (HMPV) may cause severe respiratory disease. The early innate immune response to viruses like HMPV is characterized by induction of antiviral interferons (IFNs) and proinflammatory immune mediators that are essential in shaping adaptive immune responses. Although innate immune
Estimates of individuals exposed to human metapneumovirus in a community-based Taiwanese population in 1999.
Chung-Guei Huang,Kuo-Chien Tsao,Tzou-Yien Lin,Yhu-Chering Huang,Li-Ang Lee,Tzu-Hou Chen et al.
Archives of Virology null
Analysis of the genomic sequence of a human metapneumovirus
van den Hoogen, Bernadette G, et al
Virology, 295, 119-132 (2002)
Mijia Lu et al.
Nature microbiology, 5(4), 584-598 (2020-02-06)
Internal N6-methyladenosine (m6A) modification is one of the most common and abundant modifications of RNA. However, the biological roles of viral RNA m6A remain elusive. Here, using human metapneumovirus (HMPV) as a model, we demonstrate that m6A serves as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service