Skip to Content
Merck
All Photos(1)

Key Documents

MAB5488

Sigma-Aldrich

Anti-CRABP2 Antibody

ascites fluid, Chemicon®

Synonym(s):

CRABPII

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

monoclonal

species reactivity

mouse, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CRABP2(1382)

Specificity

Reacts with Cellular Retinoic Acid Binding Protein II (CRABPII). No cross reactivity with CRABPI.

Immunogen

Full length recombinant human CRABPII.

Application

Use Anti-CRABP2 Antibody (Mouse Monoclonal Antibody) validated in ELISA, WB, ICC to detect CRABP2 also known as Cellular Retinoic Acid Binding Protein II.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shelei Pan et al.
Fluids and barriers of the CNS, 21(1), 4-4 (2024-01-09)
CSF has long been accepted to circulate throughout the subarachnoid space, which lies between the arachnoid and pia maters of the meninges. How the CSF interacts with the cellular components of the developing postnatal meninges including the dura, arachnoid, and
Yan Zheng et al.
Molecular immunology, 55(3-4), 283-291 (2013-04-04)
T cell anergy is one of the mechanisms contributing to peripheral tolerance, particularly in the context of progressively growing tumors and in tolerogenic treatments promoting allograft acceptance. We recently reported that early growth response gene 2 (Egr2) is a critical
Temporal blastemal cell gene expression analysis in the kidney reveals new Wnt and related signaling pathway genes to be essential for Wilms' tumor onset.
Maschietto, M; Trape, AP; Piccoli, FS; Ricca, TI; Dias, AA; Coudry, RA; Galante et al.
Cell Death & Disease null
Julia Derk et al.
Developmental cell, 58(8), 635-644 (2023-03-31)
The arachnoid barrier, a component of the blood-cerebrospinal fluid barrier (B-CSFB) in the meninges, is composed of epithelial-like, tight-junction-expressing cells. Unlike other central nervous system (CNS) barriers, its' developmental mechanisms and timing are largely unknown. Here, we show that mouse
Shuiliang Yu et al.
Journal of experimental & clinical cancer research : CR, 41(1), 88-88 (2022-03-10)
Resistance to standard therapy is a major reason for the poor prognosis of pancreatic ductal adenocarcinoma (PDAC). Developing novel therapy to overcome PDAC drug-resistance is urgently needed. CRABP-II was highly expressed in all PDAC but not expressed in normal pancreatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service