Skip to Content
Merck
All Photos(1)

Key Documents

MAB1683

Sigma-Aldrich

Anti-Ankyrin Antibody, clone Ank016

clone Ank016, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Ank016, monoclonal

species reactivity

rat, human, rabbit, pig, mouse, bovine, canine

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ANK1(286)

Related Categories

Specificity

Reacts with the intracellular ankyrin (216 kDa). Ankyrin consists of a single peptide of M.W. 215 kDa known to be involved in the attachment of spectrin containing membrane skeleton to the plasma membranes. Ankyrin belongs to a family of closely related polypeptides associated with the plasma membrane of cells in a variety of cell types (e.g., lymphocytes, platelets, fibroblasts and endothelial cells) and various tissues (e.g., brain, kidney, muscle and many epithelial tissues).

Immunogen

Purified human erythroid cells ankyrin

Application

Anti-Ankyrin Antibody, clone Ank016 is an antibody against Ankyrin for use in ELISA, FC, IP, WB & IC.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton
Western blotting at 1:200; low percentage PAGE gels a must; protein is quite large; proteinase inhibitors recommended or lower level fragments likely; membrane preparations and enhance signals.

ELISA: 1:1000

Immunoprecipitation: 1:100

Immunocytochemistry, using 2% paraformaldehyde fixation; higher fixation (i.e. 4% PFA can be problematic) 1:100; light fixation 5-10′ RT usually; permeabilize lightly with 0.1% triton X-100 as protein is intracellular.

Flow cytometry: 1:500

Immunohistochemistry: 1:100; light fixations suggested; PLP or mild zinc formalin recommended.

RIA: 1:5000

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Purified immunoglobulin from culture supernatant. Liquid in 10 mM sodium phosphate, pH 7.5 and 150mM NaCl with 0.01% NaN3.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P A Lang et al.
Cell death and differentiation, 12(5), 415-428 (2005-03-05)
Hyperosmotic shock, energy depletion, or removal of extracellular Cl(-) activates Ca(2+)-permeable cation channels in erythrocyte membranes. Subsequent Ca(2+) entry induces erythrocyte shrinkage and exposure of phosphatidylserine (PS) at the erythrocyte surface. PS-exposing cells are engulfed by macrophages. The present study
Najate Benamer et al.
Nature communications, 11(1), 5151-5151 (2020-10-15)
Myelination of projection neurons by oligodendrocytes is key to optimize action potential conduction over long distances. However, a large fraction of myelin enwraps the axons of parvalbumin-positive fast-spiking interneurons (FSI), exclusively involved in local cortical circuits. Whether FSI myelination contributes
Marine Ghodsi et al.
Frontiers in physiology, 14, 1205493-1205493 (2023-07-06)
Transfusion of red blood cell concentrates is the most common medical procedure to treat anaemia. However, their storage is associated with development of storage lesions, including the release of extracellular vesicles. These vesicles affect in vivo viability and functionality of
Jennifer A Tinklenberg et al.
Human molecular genetics, 27(4), 638-648 (2018-01-03)
Nemaline myopathy (NM) is a heterogeneous congenital skeletal muscle disease with cytoplasmic rod-like structures (nemaline bodies) in muscle tissue. While weakness in NM is related to contractile abnormalities, myofiber smallness is an additional abnormality in NM that may be treatable.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service