Skip to Content
Merck
All Photos(2)

Key Documents

MAB11041

Sigma-Aldrich

Anti-Serotonin receptor 1A Antibody, clone 19A9.2

clone 19A9.2, from mouse

Synonym(s):

5-hydroxytryptamine (serotonin) receptor 1A, 5-hydroxytryptamine receptor 1A, guanine nucleotide-binding regulatory protein-coupled receptor, Serotonin receptor 1A, G protein coupled receptor, 5-HT1a receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

19A9.2, monoclonal

species reactivity

human, rat

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HTR1A(3350)
rat ... Htr1A(24473)

General description

Serotonin receptor 1A, also known as 5-hydroxytryptamine receptor 1A, is a member of the G protein-coupled receptor family. Serotonin receptor 1A has been found to be expressed highly in the septum and hippocampus. Serotonin receptor 1A seems to have an inhibitory effect on Ca2+ channels, via N-channels and L-channels. The receptor actuates voltage-sensitive slowing using the N-channels and actuates steady state inhibition using the L-channels.

Immunogen

KLH-conjugated linear peptide corresponding to rat Serotonin receptor 1A.

Application

Anti-Serotonin receptor 1A Antibody, clone 19A9.2 is an antibody against Serotonin receptor 1A for use in WB, IH.
Immunohistochemistry Analysis: 1:400 dilution from a previous lot detected Serotonin receptor 1A in normal human brain tissue.
Research Category
Neuroscience
Research Sub Category
Signaling Neuroscience

Quality

Evaluated by Western Blot in rat brain tissue lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected Serotonin receptor 1A on 10 µg of rat brain tissue lysate.

Target description

~40 kDa observed. The calculated molecular weight is 46 kDa

Physical form

Format: Purified
Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emily J Reedich et al.
The Journal of physiology, 601(19), 4271-4289 (2023-08-16)
Cerebral palsy (CP) is caused by a variety of factors that damage the developing central nervous system. Impaired motor control, including muscle stiffness and spasticity, is the hallmark of spastic CP. Rabbits that experience hypoxic-ischaemic (HI) injury in utero (at
Laura García-Durán et al.
International journal of molecular sciences, 22(19) (2021-10-14)
Selective 5-HT reuptake inhibitor antidepressants (SSRIs) are the first choice in major depressive disorder (MDD), but 50% of affected patients do not show improvement. Galanin(1-15) [GAL(1-15)] enhanced Fluoxetine antidepressant-like effects in an animal model of depression, the olfactory bulbectomy (OBX);
Shaoyuan Li et al.
Molecular pain, 14, 1744806918787368-1744806918787368 (2018-06-21)
Painful neuropathy is a frequent comorbidity in diabetes. Zucker diabetic fatty (fa/fa) rats develop type 2 diabetes spontaneously with aging and show nociceptive hypersensitivity at the age of 13 weeks. In preclinical and clinical studies, the treatment of diabetic neuropathy
Elenia Cinelli et al.
The European journal of neuroscience, 52(8), 3903-3917 (2020-05-08)
In mammals, 5-HTexcitatory respiratory effects imply 5-HT1A receptor-mediated disinhibition of pre-Bötzinger complex neurons. In the lamprey, 5-HT1A receptors are involved in the neural control of locomotion, but their role in the respiratory regulation, particularly at the level of the putative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service