LUXGCS
mPAGE® Lux Curing Station
Synonym(s):
gel curing instrument
About This Item
Recommended Products
AC/DC input
100 - 240 V, 1.5 A
description
Instrument for curing mini gels using UV light, does not include casters, plates or combs.
packaging
pkg of 1 pack
manufacturer/tradename
mPAGE®
greener alternative product characteristics
Less Hazardous Chemical Syntheses
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
parameter
15-30 °C ambient temp. limit
20-80% humidity limit (non-condensing)
technique(s)
SDS-PAGE: suitable
electrophoresis: suitable
H
>2000 m , altitude
L × W × H
41.275 cm × 29.21 cm × 32.385 cm
gel size
8.3 cm × 10.1 cm
gel thickness
0.75 mm
1.0 mm
1.5 mm
well size × volume
0.75 mm × 10 well (25 μL) × 15 well (10 μL)
1.0 mm × 10 well (40 μL) × 15 well (25 μL)
1.5 mm × 10 well (60 μL) × 15 well (36 μL)
well volume
10 well , using 10-well comb
15 well , using 15-well comb
greener alternative category
General description
Components
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Refresh your gel casting workflow with ready-to-use SDS-PAGE gels in just 3 minutes. Discover faster, simpler, and safer gel casting methods.
Refresh your gel casting workflow with ready-to-use SDS-PAGE gels in just 3 minutes. Discover faster, simpler, and safer gel casting methods.
Refresh your gel casting workflow with ready-to-use SDS-PAGE gels in just 3 minutes. Discover faster, simpler, and safer gel casting methods.
Refresh your gel casting workflow with ready-to-use SDS-PAGE gels in just 3 minutes. Discover faster, simpler, and safer gel casting methods.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service