Skip to Content
Merck
All Photos(1)

Key Documents

CBL1366

Sigma-Aldrich

Anti-CD140a Antibody, clone APA5

clone APA5, Chemicon®, from rat

Synonym(s):

PDGF Receptor alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

APA5, monoclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PDGFRA(5156)

Specificity

Specifically recognizes mouse CD140a, the alpha chain of PDGF receptor. A receptor tyrosine kinase, PDGFRa forms dimers on the surface upon ligand binding and phosphorylates substrates. CD140a is expressed by embryonic tissues and mesenchymal-derived cells of the adult mouse tissues.

Application

Detect CD140a using this Anti-CD140a Antibody, clone APA5 validated for use in ELISA, FC, WB, IH.
Flow Cytometry: ≤ 1 μg per 10E6 cells

ELISA

Immunohistochemistry: 10μg/mL

Blocking of Ligand Binding

Western Blotting

Optimal working dilutions must be determined by the end user.
Research Category
Signaling
Research Sub Category
Growth Factors & Receptors

Physical form

Format: Purified
Purified immunoglobulin in PBS, 150 mM NaCl, pH 7.2, containing 0.09% sodium azide.

Storage and Stability

Maintain at 2-8°C for up to 12 months from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Anesti et al.
Cells, 11(11) (2022-06-11)
Two main stem cell pools exist in the postnatal mammalian brain that, although they share some "stemness" properties, also exhibit significant differences. Multipotent neural stem cells survive within specialized microenvironments, called niches, and they are vulnerable to ageing. Oligodendroglial lineage-restricted
Magali Macchi et al.
eLife, 9 (2020-06-10)
Myelin destruction is followed by resident glia activation and mobilization of endogenous progenitors (OPC) which participate in myelin repair. Here we show that in response to demyelination, mature oligodendrocytes (OLG) bordering the lesion express Ndst1, a key enzyme for heparan
Bilal El Waly et al.
iScience, 25(10), 105102-105102 (2022-10-04)
After demyelinating insult, the neuronal progenitors of the adult mouse sub-ventricular zone (SVZ) called neuroblasts convert into oligodendrocytes that participate to the remyelination process. We use this rare example of spontaneous fate conversion to identify the molecular mechanisms governing these
Haley E Titus et al.
Glia, 70(10), 1950-1970 (2022-07-10)
Multiple sclerosis (MS) is a central nervous system (CNS) autoimmune disease characterized by inflammation, demyelination, and neurodegeneration. The ideal MS therapy would both specifically inhibit the underlying autoimmune response and promote repair/regeneration of myelin as well as maintenance of axonal
Lida Zoupi et al.
Glia, 66(3), 576-591 (2017-11-23)
The oligodendrocyte maturation process and the transition from the pre-myelinating to the myelinating state are extremely important during development and in pathology. In the present study, we have investigated the role of the cell adhesion molecule CNTN2/TAG-1 on oligodendrocyte proliferation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service