Skip to Content
Merck
All Photos(1)

Key Documents

AP189P

Sigma-Aldrich

Donkey Anti-Rat IgG Antibody, HRP conjugate, Species Adsorbed

Chemicon®, from donkey

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

donkey

Quality Level

conjugate

peroxidase conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Application

Donkey anti-Rat IgG Antibody, HRP conjugate, Species Adsorbed detects level of Donkey Rat IgG & has been published & validated for use in ELISA & WB.
Research Category
Secondary & Control Antibodies
Research Sub Category
Secondary Antibodies Adsorbed for Dual Labeling

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaodong Peng et al.
Oncology reports, 40(1), 405-413 (2018-05-17)
The roles and model of action of the chromatin assembly complex factor-1B (CHAF1B) gene in liver cancer have not been fully elucidated. The CHAF1B gene in human hepatocellular carcinoma cell line HUH-7 was knocked down using a lentivirus and the
Ryuichi Kimura et al.
Developmental dynamics : an official publication of the American Association of Anatomists, 251(3), 525-535 (2021-09-21)
Repressor element 1-silencing transcription factor (REST) is a master regulator that is highly expressed in multipotent stem cells to repress gene networks involving a wide range of biological processes. A recent study has suggested that REST might be involved in
Natasha K Brockwell et al.
NPJ precision oncology, 3, 21-21 (2019-09-05)
Patients diagnosed with triple negative breast cancer (TNBC) have an increased risk of rapid metastasis compared to other subtypes. Predicting long-term survival post-chemotherapy in patients with TNBC is difficult, yet enhanced infiltration of tumor infiltrating lymphocytes (TILs) has been associated
Taifeng Zhou et al.
eLife, 9 (2020-03-19)
Mechanical forces are fundamental regulators of cell behaviors. However, molecular regulation of mechanotransduction remain poorly understood. Here, we identified the mechanosensitive channels Piezo1 and Piezo2 as key force sensors required for bone development and osteoblast differentiation. Loss of Piezo1, or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service