Skip to Content

AM76

Anti-TBK1 Mouse mAb (108A429)

liquid, clone 108A429, Calbiochem®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.43
UNSPSC Code:
12352203
Clone:
108A429, monoclonal
Species reactivity:
human
Application:
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

antibody form

purified antibody

antibody product type

primary antibodies

clone

108A429, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

dilution

(Immunoblotting (2 µg/mL))

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TBK1(29110)

General description

Protein G purified mouse monoclonal antibody. Recognizes the ~80 kDa TBK1 protein.
Recognizes the ~80 kDa TBK1 protein in HEK293 cells.
This Anti-TBK1 Mouse mAb (108A429) is validated for use in Immunoblotting for the detection of TBK1.

Immunogen

Human
full-length, recombinant, human TBK1 fused to a His•Tag sequence

Application



Immunoblotting (2 g/ml)

Packaging

Please refer to vial label for lot-specific concentration.

Physical form

In PBS, 0.2% gelatin.

Preparation Note

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Negative Control
Normal mouse serum
Positive Control
HEK293 cells

Other Notes

Antibody should be titrated for optimal results in an individual systems.
Pomeerantz, J.L. and Baltimore, D. 1999. EMBO J.18, 6694.
Didonato, J.A., et al. 1997. Nature388, 548.
Lee, F.S., et al. 1997. Cell88, 213.
Malinin, N.L., et al. 1997. Nature385, 540.
Mercurio, F., et al. 1997. Science278, 860.
Regnier, C.H., et al. 1997. Cell90, 373.
Verma, I. and Stevenson, J.K. 1997. Proc. Natl. Acad. Sci. USA94, 11758.
Verma, I.M., et al. 1995. Genes Dev.9, 2723.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library