Skip to Content
Merck

ABC102

Anti-CCN1/CYR61 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Protein CYR61, CCN family member 1, Cysteine-rich angiogenic inducer 61, Insulin-like growth factor-binding protein 10, IBP-10, IGF-binding protein 10, IGFBP-10, Protein GIG1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
4
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, rat

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CYR61(3491)

General description

CCN1 (cysteine-rich 61 or CYR61), a ubiquitous angiogenic factor, is a member of the CNN family, which is also comprised of CTGF, WISP-1, WISP-2, and WISP-3. It is a multidomain protein, containing one each of CTCK, IGFBP, TSP type-1, and VWFC domains. CCN1 is associated with the extracellular matrix and functions as a ligand for integrin receptors. It is expressed in response to growth factor stimulation, and mediates various cellular processes, including cell adhesion, migration, proliferation, angiogenesis, extracellular-matrix remodeling, and placental development. Previous studies have indicated that it may also play a role in wound healing by influencing the expression of genes required for this process, such as TIMP1, PAI-1, VEGA-A, VEGA-C, MMP1, MMP3, and uPA. CCN1 has also been implicated in breast cancer.
~53 kDa observed. The calculated molecular weight of this protein is 42 kDa, but can be observed at higher molecular weights due to glycosylation. A degradation fragment may be observed at ~15 kDa in some lysates. (Rother et al., 2010)

Immunogen

Epitope: CTCK domain
KLH-conjugated linear peptide corresponding to the CTCK domain of human CCN1/CYR61.

Application

Detect CCN1/CYR61 using this Anti-CCN1/CYR61 Antibody validated for use in Western Blotting, IHC(P).
Immunohistochemistry Analysis: 1:200 dilution from a representative lot detected CCN1/CYR61 in human kidney tissue.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Biochem/physiol Actions

This antibody recognizes the CTCK domain of: CCN1/CYR61

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
NIH/3T3 cell lysate
Evaluated by Western Blot in NIH/3T3 cell lysate.

Western Blot Analysis: 2 µg/mL of this antibody detected CCN1/CYR61 in 10 µg of NIH/3T3 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library