Skip to Content
Merck
All Photos(1)

Key Documents

AB125

Sigma-Aldrich

Anti-Serotonin Antibody

serum, Chemicon®

Synonym(s):

5-HT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

shipped in

dry ice

target post-translational modification

unmodified

Specificity

Serotonin (5-Hydroxytryptamine)

The cross-reactivities were determined using an ELISA test by competition experiments with the following compounds:



Compound Cross-reactivity

5-Hydroxytryptamine-G-PL 1

5-Methoxytryptamine-G-PL 1/25

Tryptamine-G-PL 1/60

5-Hydroxytryptophan-G-PL 1/175

5-Hydroxytryptamine 1/1150

Tryptophan-G-PL 1/3400

5-Methoxytryptophan-G-PL 1/19800



The antisera was also tested for specificity using the free-floating PAP technique on rat

raphe nuclei.



Abbreviations:



(G) Glutaraldehyde

(=) Non-reduced conjugate

(PL) Poly-lysine

Immunogen

5-Hydroxytryptamine-glutaraldehyde-Poly-lysine

Application

Anti-Serotonin Antibody detects level of Serotonin & has been published & validated for use in IH.
Immunohistochemistry: 1:1,000-1:2,500 by ABC or enhanced detection. Glutaraldehyde fixation is required. Antibody will not react with non-glutaraldehyde treated tissues. Blocking should be with serum or BSA containing 0.1-0.2% triton X-100; remove all blocking an detergents during primary antibody incubation. Dilute only in PBS or TBS for best results. For fluorescent detection antibody dilution must be reduced, but it is not recommended because of glutaraldehyde autofluorescence.



Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Physical form

Rabbit antiserum. Liquid with 0.05% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Lu et al.
The Journal of comparative neurology, 508(4), 648-662 (2008-04-03)
Classical anesthetics of the gamma-aminobutyric acid type A receptor (GABA(A))-enhancing class (e.g., pentobarbital, chloral hydrate, muscimol, and ethanol) produce analgesia and unconsciousness (sedation). Dissociative anesthetics that antagonize the N-methyl-D-aspartate (NMDA) receptor (e.g., ketamine, MK-801, dextromethorphan, and phencyclidine) produce analgesia but
Katia Carneiro et al.
BMC developmental biology, 11, 29-29 (2011-05-24)
Consistent asymmetry of the left-right (LR) axis is a crucial aspect of vertebrate embryogenesis. Asymmetric gene expression of the TGFβ superfamily member Nodal related 1 (Nr1) in the left lateral mesoderm plate is a highly conserved step regulating the situs
Shih-Chang Hsueh et al.
International journal of molecular sciences, 19(2) (2018-02-09)
Parkinson's disease (PD) is typically characterized by impairment of motor function. Gait disturbances similar to those observed in patients with PD can be observed in animals after injection of neurotoxin 6-hydroxydopamine (6-OHDA) to induce unilateral nigrostriatal dopamine depletion. Exercise has
Sujan Gautam et al.
iScience, 27(1), 108616-108616 (2024-01-05)
Studies across a diverse group of metazoan embryos indicate that Wnt signaling often activates the transcription factor Sp5, forming a signaling 'cassette' that plays critical roles in many developmental processes. This study explores the role of Wnt/Sp5 signaling during the
Robert A Kozol et al.
eLife, 12 (2023-07-27)
The vertebrate brain is highly conserved topologically, but less is known about neuroanatomical variation between individual brain regions. Neuroanatomical variation at the regional level is hypothesized to provide functional expansion, building upon ancestral anatomy needed for basic functions. Classically, animal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service