8.56193
Fmoc-Lys(biotinyl-ε-aminocaproyl)-NovaSyn® TGR A resin
Novabiochem®
About This Item
Recommended Products
Quality Level
product line
NovaSyn® TG
Novabiochem®
form
beads
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
manufacturer/tradename
Novabiochem®
application(s)
peptide synthesis
functional group
Fmoc
storage temp.
2-8°C
Related Categories
General description
Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Biotin-labelled peptides find applications in immunology and histochemistry for affinity purification and receptor localization.
Biotin-labelled peptides find applications in immunology and histochemistry for affinity purification and receptor localization.
Biotin-labelled peptides find applications in immunology and histochemistry for affinity purification and receptor localization.
Biotin-labelled peptides find applications in immunology and histochemistry for affinity purification and receptor localization.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service