Skip to Content
Merck
All Photos(1)

Key Documents

8.52059

Sigma-Aldrich

Fmoc-Hse(Trt)-OH

Novabiochem®

Synonym(s):

Fmoc-Hse(Trt)-OH, N-α-Fmoc-O-trityl-L-homoserine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H33NO5
CAS Number:
Molecular Weight:
583.67
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

Quality Level

product line

Novabiochem®

Assay

≥80.0% (acidimetric)
≥98% (TLC)
≥98.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

hydroxyl

storage temp.

15-25°C

SMILES string

N([C@@H](CCOC(c6ccccc6)(c5ccccc5)c4ccccc4)C(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C38H33NO5/c40-36(41)35(39-37(42)43-26-34-32-22-12-10-20-30(32)31-21-11-13-23-33(31)34)24-25-44-38(27-14-4-1-5-15-27,28-16-6-2-7-17-28)29-18-8-3-9-19-29/h1-23,34-35H,24-26H2,(H,39,42)(H,40,41)/t35-/m0/s1

InChI key

QUTREFXHXPNEJN-DHUJRADRSA-N

General description

Building block which enables side-chain modification of homoserine serines by Fmoc SPPS. The Trt group can be removed with 1% TFA in DCM containing 5% TIS, enabling the side-chain hydroxyl group to be selectively modified whilst the derivative is attached to the solid support. This derivative has been employed to prepare oligonucleotide conjugates [1].

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS


Literature references

[1] D. A. Stetsenko & M. J. Gait (2001) Bioconjugate Chem. , 12, 576.

Linkage

Replaces: 04-12-1126

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Assay (HPLC, area%): ≥ 98.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble
Assay (acidimetric): ≥ 80.0 %
Water (K. F.): ≤ 1.00 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Novabiochem® offers orthogonally protected amino acids for peptide synthesis, including cyclic and branched peptides.

Novabiochem® offers orthogonally protected amino acids for peptide synthesis, including cyclic and branched peptides.

Novabiochem® offers orthogonally protected amino acids for peptide synthesis, including cyclic and branched peptides.

Novabiochem® offers orthogonally protected amino acids for peptide synthesis, including cyclic and branched peptides.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service