Skip to Content
Merck
All Photos(1)

Key Documents

8.43803

Sigma-Aldrich

2-Methoxy-4-vinylphenol

for synthesis

Synonym(s):

2-Methoxy-4-vinylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O2
CAS Number:
Molecular Weight:
150.17
MDL number:
UNSPSC Code:
12352104
EC Index Number:
232-101-2
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥96.0% (GC)

bp

224 °C/1013 hPa

mp

9-10 °C

density

1.11 g/cm3 at 14 °C

storage temp.

2-30°C

SMILES string

O(C)c1c(ccc(c1)C=C)O

InChI

1S/C9H10O2/c1-3-7-4-5-8(10)9(6-7)11-2/h3-6,10H,1H2,2H3

InChI key

YOMSJEATGXXYPX-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 96.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.106 - 1.110
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service