Skip to Content
Merck
All Photos(1)

Key Documents

8.20966

Sigma-Aldrich

n-Pentyl nitrite

for synthesis

Synonym(s):

n-Pentyl nitrite, n-Amyl nitrite, Nitrous acid n-pentyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4ONO
CAS Number:
Molecular Weight:
117.15
UNSPSC Code:
12352108
EC Index Number:
207-332-7
NACRES:
NA.22

vapor pressure

65 hPa ( 20 °C)

Quality Level

Assay

≥95.0% (GC)

form

liquid

autoignition temp.

205 °C

bp

104 °C/1013 hPa

transition temp

flash point -40 °C

density

0.88 g/cm3 at 20 °C

storage temp.

2-8°C

InChI

1S/C5H11NO2/c1-2-3-4-5-8-6-7/h2-5H2,1H3

InChI key

CSDTZUBPSYWZDX-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.879 - 0.883
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

32.0 °F

Flash Point(C)

0 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service