Skip to Content
Merck
All Photos(1)

Key Documents

8.20155

Sigma-Aldrich

2-Biphenylylamine

for synthesis

Synonym(s):

2-Biphenylylamine, 2-Aminodiphenyl, 2-Phenylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4-2-(NH2)
CAS Number:
Molecular Weight:
169.22
MDL number:
UNSPSC Code:
12352116
EC Index Number:
201-990-9
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

form

powder

autoignition temp.

461 °C

potency

2340 mg/kg LD50, oral (Rat)

bp

298-300 °C

mp

46-48 °C

transition temp

flash point 153 °C

storage temp.

2-30°C

SMILES string

Nc1c(cccc1)c2ccccc2

InChI

1S/C12H11N/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-9H,13H2

InChI key

TWBPWBPGNQWFSJ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 46 °C
Melting range (upper value): ≤ 50 °C
p-Isomeres (GC, area%): ≤ 0.10 % (a/a)
Identity (IR): passes test

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

307.4 °F - closed cup

Flash Point(C)

153 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service