Skip to Content
Merck
All Photos(1)

Key Documents

8.08616

Sigma-Aldrich

Taurine

for peptide synthesis

Synonym(s):

Taurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H7NO3S
CAS Number:
Molecular Weight:
125.15
MDL number:
UNSPSC Code:
12352106
EC Index Number:
203-483-8
NACRES:
NA.22

Product Name

Taurine, for synthesis

Quality Level

form

powder

potency

>5000 mg/kg LD50, oral (Rat)

mp

305 °C (decomposition)

solubility

79 g/L

density

1.72 g/cm3 at 20.1 °C

storage temp.

no temp limit

SMILES string

[S](=O)(=O)([O-])CC[N+H3]

InChI

1S/C2H7NO3S/c3-1-2-7(4,5)6/h1-3H2,(H,4,5,6)

InChI key

XOAAWQZATWQOTB-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service