8.05967
3-Mercaptopropionic acid
for synthesis
Synonym(s):
3-Mercaptopropionic acid
About This Item
Recommended Products
vapor pressure
0.053 hPa ( 20 °C)
Quality Level
Assay
≥98.0% (GC)
form
liquid
autoignition temp.
350 °C
potency
96 mg/kg LD50, oral (Rat)
expl. lim.
1.60 % (v/v)
pH
2 (20 °C, 120 g/L in H2O)
bp
110 °C/20 hPa
mp
17-19 °C
transition temp
flash point 93 °C
density
1.22 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
SCCC(=O)O
InChI
1S/C3H6O2S/c4-3(5)1-2-6/h6H,1-2H2,(H,4,5)
InChI key
DKIDEFUBRARXTE-UHFFFAOYSA-N
Application
- Thiol-ene click cross-linking for material science: Research on thiol-ene click cross-linking of starch oleate films investigated that 3-Mercaptopropionic acid is used to esterified a series of polyethylene glycol (PEGs) with varying molecular weights during the synthesis of biobased films (Boetje et al., 2023).
- Redox-responsive nanoparticle systems for cancer therapy: A study utilized 3-Mercaptopropionic acid in the synthesis of gold nanoparticles coated with hyaluronic acid and folic acid, demonstrating its application in targeted anticancer therapy through a redox-responsive mechanism (Munderere et al., 2024).
- Preparation of superhydrophobic materials: The application of 3-Mercaptopropionic acid in the preparation of superhydrophobic fabric via Michael addition under ultraviolet light was explored, indicating its utility in creating water-resistant textile surfaces (Wang et al., 2023).
- Optoelectronic applications in nanocomposite films: Research involved 3-Mercaptopropionic acid in the eco-friendly electrophoretic deposition of fluorescent nanocomposite films, employing quantum dots for enhanced optoelectronic applications, reflecting its role in sustainable materials engineering (Morimoto et al., 2024).
Analysis Note
Density (d 20 °C/ 4 °C): 1.220 - 1.222
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B
Storage Class Code
6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 1
Flash Point(F)
200.1 °F - closed cup
Flash Point(C)
93.4 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service