Skip to Content
Merck
All Photos(1)

Key Documents

71078

Millipore

RedAlert 10X Western Blot Stain

Visualize proteins transferred to nitrocellulose or PVDF membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116133
NACRES:
NB.23

form

liquid

manufacturer/tradename

Novagen®

storage condition

OK to freeze

shipped in

ambient

storage temp.

10-30°C

General description

RedAlert stain is used to visualize proteintransferred to membranes prior to Westernblot analysis or protein sequencing by Edmandegradation. RedAlert stain binds reversiblyto proteins on nitrocellulose and PVDFmembranes and can also stain proteins boundto cellulose acetate. The RedAlert Stain bindspositively charged amino groups and nonpolarregions of proteins, resulting in reddish-pinkbands. This stain can be used to verify transferof proteins before immunological detectionon nitrocellulose or PVDF membranes orto detect blood serum proteins on celluloseacetate media. Because the stain is reversible,it is a good choice for detecting proteins priorto protein sequencing or HPLC analysis. Thesensitivity of the stain is approximately 50 ngof protein. The RedAlert Stain is provided as a10X solution.

Warning

Toxicity: Irritant (B)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service