Skip to Content
Merck
All Photos(1)

Key Documents

554720

Millipore

trans-Retinoic Acid

Potent modulator of growth and differentiation. Inhibits melanocyte adhesion, motility, and growth.

Synonym(s):

trans-Retinoic Acid, Tretinoin, ATRA, Vitamin A Acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28O2
CAS Number:
Molecular Weight:
300.44
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.21
Pricing and availability is not currently available.

description

Merck USA index - 14, 8165

Quality Level

Assay

≥95% (by assay)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

yellow

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

OC(=O)\C=C(/C)\C=C\C=C(/C)\C=C\C1=C(CCCC1(C)C)C

InChI

1S/C20H28O2/c1-15(8-6-9-16(2)14-19(21)22)11-12-18-17(3)10-7-13-20(18,4)5/h6,8-9,11-12,14H,7,10,13H2,1-5H3,(H,21,22)/b9-6+,12-11+,15-8+,16-14+

InChI key

SHGAZHPCJJPHSC-YCNIQYBTSA-N

General description

Potent modulator of growth and differentiation. Exerts its effects by binding to nuclear retinoic acid receptors (RARs) which directly regulate gene expression. Inhibits melanocyte adhesion, motility, and growth. Has keratolytic activity. Induces differentiation of tumor cells of neural origin. Induces apoptosis in acute promyelocytic carcinoma cells. PROTECT FROM AIR.

Application


  • All-trans-retinoic acid modulates glycolysis via H19 and telomerase: the role of mir-let-7a in estrogen receptor-positive breast cancer cells.: This research reveals how all-trans-retinoic acid influences glycolysis in breast cancer cells by modulating H19 and telomerase, demonstrating its potential in breast cancer therapy (El Habre et al., 2024).

  • Retinoic acid tiers mitochondrial metabolism to Sertoli Cell-Mediated efferocytosis via a non-RAR-dependent mechanism.: The study explores the role of retinoic acid in linking mitochondrial metabolism to efferocytosis in Sertoli cells, providing insights into its non-RAR-dependent mechanisms and potential applications in reproductive biology (Wu et al., 2024).

  • Combined treatment of All-trans retinoic acid with Tamoxifen suppresses ovarian cancer.: This article discusses the synergistic effects of combining all-trans-retinoic acid with tamoxifen in suppressing ovarian cancer, highlighting a promising therapeutic strategy for ovarian cancer patients (Xu et al., 2024).

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). DMSO stock solutions are stable for up to 2 weeks at 4°C.

Other Notes

Tosi, P., et al. 1994. Leuk. Lymphoma14, 503.
Clagett-Dame, M., et al. 1993. Arch. Biochem. Biophys.300, 684.
Labbaye, C., et al. 1993. Blood81, 475.
Sakashita, A., et al. 1993. Blood81, 1009.
Situ, R., et al. 1993. Dermatology186, 38.
Tini, M., et al. 1993. Genes Develop.7, 295.
Leid, M., et al. 1992. Trends Biochem. Sci.17, 427.
Sharpe, C.R. 1991. Neuron7, 239.
Thaller, C. and Eichele, G. 1990. Nature345, 815.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natalia Moskal et al.
Nature communications, 11(1), 88-88 (2020-01-05)
The accumulation of damaged mitochondria causes the death of dopaminergic neurons. The Parkin-mediated mitophagy pathway functions to remove these mitochondria from cells. Targeting this pathway represents a therapeutic strategy for several neurodegenerative diseases, most notably Parkinson's disease. We describe a

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Can I reconstitute this product then aliquot and store at -20C for extended periods of time?

    1 answer
    1. Freezing is acceptable. A stock solution of 0.01 M (3 mg/ml) Retinoic acid in absolute ethanol was stored at -70°C for up to two weeks, according to Ponzoni and Lanciotti in Neurochemistry, volume 54, page 540, 1990.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service