Skip to Content
Merck
All Photos(1)

Key Documents

552233

Sigma-Aldrich

QX-314

≥99% (HPLC), lyophilized solid, voltage-sensitive Na+ conductance blocker, Calbiochem®

Synonym(s):

QX-314

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H27BrN2O
CAS Number:
Molecular Weight:
343.30
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

QX-314, A quaternary lidocaine derivative, permanently charged, and lipophobic.

Quality Level

Assay

≥99%

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

solubility

water: 3 mg/mL

shipped in

ambient

storage temp.

10-30°C

SMILES string

[N+](CC)(CC)(CC)CC(=O)Nc1c(cccc1C)C

InChI

1S/C16H26N2O/c1-6-18(7-2,8-3)12-15(19)17-16-13(4)10-9-11-14(16)5/h9-11H,6-8,12H2,1-5H3/p+1

InChI key

PYEBKOFMWAMBFV-UHFFFAOYSA-O

General description

A quaternary lidocaine derivative, permanently charged, and lipophobic. A powerful blocker of voltage-sensitive Na+ conductance when applied intracellularly. Suppresses the generation of Na+-dependent spikes from inside the cell membrane, without affecting Ca2+ currents or glutamate-activated currents.

Biochem/physiol Actions

Cell permeable: no
Primary Target
A powerful blocker of voltage-sensitive Na+ conductance
Product does not compete with ATP.
Reversible: no
fully active in neurophysiological testing

Warning

Toxicity: Irritant (B)

Other Notes

Mooney, R. 1992. J. Neurosci.12, 2464.
Regehr, W.G. and Tank, D.W. 1992. J. Neurosci.12, 4202.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael G Frantz et al.
Current biology : CB, 30(15), 2962-2973 (2020-06-27)
Disrupting binocular vision during a developmental critical period can yield enduring changes to ocular dominance (OD) in primary visual cortex (V1). Here we investigated how this experience-dependent plasticity is coordinated within the laminar circuitry of V1 by deleting separately in
Charlie H T Kwok et al.
Frontiers in immunology, 12, 626884-626884 (2021-04-27)
Increased afferent input resulting from painful injury augments the activity of central nociceptive circuits via both neuron-neuron and neuron-glia interactions. Microglia, resident immune cells of the central nervous system (CNS), play a crucial role in the pathogenesis of chronic pain.
Emmanuel L Crespo et al.
STAR protocols, 2(3), 100667-100667 (2021-07-22)
Bioluminescent optogenetics (BL-OG) allows activation of photosensory proteins, such as opsins, by either fiberoptics or by administering a luciferin. BL-OG thus confers both optogenetic and chemogenetic access within the same genetically targeted neuron. This bimodality offers a powerful approach for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service