Skip to Content
Merck
All Photos(1)

Key Documents

5.42765

Millipore

Enrichment Sample Homogenizer (ESH)

sample homogenizer, suitable for sample preparation

Synonym(s):

Sample mixer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41101504
NACRES:
NA.84

application(s)

food and beverages
pathogen testing

sample preparation

storage temp.

2-30°C

General description

Homogenize food samples with confidence prior to microbiological and molecular analysis with the easy-to-use Enrichment Sample Homogenizer (ESH). The ESH instrument is used for the preparation of solid samples before analysis. The ESH instrument does not damage potential pathogenic bacteria contained in the sample and does not require sterilization. This reliable and powerful Homogenizer enables rapid and efficient sample homogenization with quiet operation. The Multi-function digital display/control panel, Adjustable blending speed (4, 6, 8, and 10 strokes) and Variable time (1 second to 1 hour or open running/countdown) allow users to adapt to any sample type. This instrument weighs approximately 16 kilograms.

Application

The Enrichment Sample Homogenizer is used for automatic homogenization of food enrichment samples with massaging paddles.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Updates on ISO standards in food microbiology by expert Barbara Gerten from MilliporeSigma

Updates on ISO standards in food microbiology by expert Barbara Gerten from Merck

Updates on ISO standards in food microbiology by expert Barbara Gerten from Merck

Updates on ISO standards in food microbiology by expert Barbara Gerten from Merck

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service