Skip to Content
Merck
All Photos(1)

Key Documents

527621

Sigma-Aldrich

Plasmin, Human Plasma

Synonym(s):

Plasmin, Human Plasma

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

description

Merck USA index - 14, 7520

Quality Level

form

liquid

specific activity

≥5 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−20°C

General description

Native plasmin from human plasma. Catalyzes the hydrolysis of peptides, arginine and lysine esters and converts fibrin into soluble products.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will hydrolyze 1.0 µmol of N-tosyl-Gly-Pro-Lys-pNA per min at 25°C, pH 7.8.

Physical form

In 100 mM sodium phosphate, 1 mM 6-aminohexanoic acid, 25% glycerol, pH 7.3.

Preparation Note

Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Federici, A.B., et al. 1993. Blood 81, 720.
Hoffmann, J.J., and Janssen, W.C. 1992. Thromb. Res. 67, 711.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J J Hoffmann et al.
Thrombosis research, 67(6), 711-719 (1992-09-15)
The mechanisms by which thrombolytic agents affect platelet function are not yet elucidated. The aim of the present study was to investigate the effects of plasmin, generated by thrombolytic agents in plasma, on platelet glycoproteins (GP) Ib and IIb/IIIa. Platelet-rich
A B Federici et al.
Blood, 81(3), 720-725 (1993-02-01)
The behavior of plasma von Willebrand factor (vWF) in patients with acute leukemia (n = 5), decompensated cirrhosis (n = 10), and acute pancreatitis (n = 5) was investigated to evaluate whether the systemic proteolytic states associated with these diseases

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service