Skip to Content
Merck
All Photos(1)

Key Documents

5.00511

Sigma-Aldrich

Histone Acetyltransferase Inhibitor VIII, NU9056

Synonym(s):

Histone Acetyltransferase Inhibitor VIII, NU9056, HAT Inhibitor VIII, p300/CBP Inhibitor VII, PCAF Inhibitor V, 1,2-Bis(isothiazol-5-yl)disulfane, 1,2-di(isothiazol-5-yl)disulfane, KAT5 Inhibitor I, Tip60 Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4N2S4
CAS Number:
Molecular Weight:
232.37
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥97% (HPLC)

Quality Level

form

oil

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow-brown

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

[s]1nccc1SSc2[s]ncc2

InChI

1S/C6H4N2S4/c1-3-7-9-5(1)11-12-6-2-4-8-10-6/h1-4H

InChI key

MLRAMCAHIWHWRM-UHFFFAOYSA-N

General description

NU9056, a disulfane compound, is reported to be a potent inhibitor of lysine acetyltransferase 5 (KAT5). NU9056 inhibits protein acetylation in prostate cancer cell lines. It acts against histone acetyltransferase KAT5/Tip60 than PCAF, p300, and GCN5 in cell-free HAT activity assays (IC50 ≤2, ≤36, ≤60, and >100 µM, respectively) and effectively reduces H3K14, H4K8, H4K16, but not α-tubulin, acetylation level in LNCaP prostate cancer cells (max inhibition at 5-10 µM). Shown to induce cell cycle arrest and apoptosis (17 to 24 µM) in LNCaP cultures and exhibit antiproliferation activity in both androgen-responsive and androgen-independent CaP cultures (GI50 from 7.5 to 27 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
KAT5
Reversible: yes

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenjing Xu et al.
International journal of endocrinology, 2022, 2014568-2014568 (2022-02-11)
Anaplastic thyroid carcinoma (ATC) is considered to be one of the most aggressive cancers. Our previous study proved that highly expressed lysine acetyltransferase 5 (KAT5) in ATC is associated with a poorer prognosis. Here, this study examined the effects of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service