499635
OSC Inhibitor, Ro 48-8071
The OSC Inhibitor, Ro 48-8071, also referenced under CAS 189197-69-1, controls the biological activity of 2,3-oxidosqualene:lanosterol cyclase activity (OSC).
Synonym(s):
OSC Inhibitor, Ro 48-8071, 4ʹ-(6-(Allylmethylamino)hexyloxy)-4-bromo-2ʹ-fluorobenzophenone, fumarate
About This Item
Recommended Products
Quality Level
Assay
≥95% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated (hygroscopic)
protect from light
color
off-white
solubility
water: 5 mg/mL
shipped in
ambient
storage temp.
2-8°C
SMILES string
Fc1c(ccc(c1)OCCCCCCN(CC=C)C)C(=O)c2ccc(cc2)Br
InChI
1S/C23H27BrFNO2/c1-3-14-26(2)15-6-4-5-7-16-28-20-12-13-21(22(25)17-20)23(27)18-8-10-19(24)11-9-18/h3,8-13,17H,1,4-7,14-16H2,2H3
InChI key
CMYCCJYVZIMDFU-UHFFFAOYSA-N
General description
Warning
Reconstitution
Other Notes
Peffley, D.M., et al. 1998. Biochem. Pharmacol.56, 439.
Morand, O.H., et al. 1997. J. Lipid Res.38, 373.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service