Skip to Content
Merck
All Photos(1)

Key Documents

371754

Sigma-Aldrich

Anti-Gqα- and G₁₁α-Subunits, Internal (283-300) Rabbit pAb

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

does not contain

preservative

species reactivity

human, mouse, rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−70°C

target post-translational modification

unmodified

Gene Information

human ... GNAI1(2770)
mouse ... Gnai1(14677)
rat ... Gnai1(25686)

General description

Anti-Gqα- and G₁₁α-Subunits, Internal (283-300), rabbit polyclonal, recognizes the ~42 kDa Gqα- and the ~43 kDa G₁₁α-subunits. Validated for use in WB and neutralization studies.
Protein A purified rabbit polyclonal antibody. Recognizes the ~42 kDa Gqα- and the ~43 kDa G11α-subunit proteins.
Recognizes the ~42 kDa Gqα- and the ~43 kDa G11α-subunits. Does not cross-react with other G-proteins of the non-pertussis toxin-sensitive family.

Immunogen

a synthetic peptide (IMYSHLVDYFPEYDGPQR) corresponding to an internal amino acid sequence shared by human, mouse, and rat Gqα- and G₁₁α-subunits

Application

Immunoblotting (1:1000)

Neutralization Studies (see comments)

Warning

Toxicity: Standard Handling (A)

Other Notes

The antibody recognizes both Gqα and G11α since they both share the internal sequence represented by the immunizing peptide. The antibody may also be useful to block receptor G protein coupling. Does not cross-react with other G proteins of the non-pertussis toxin sensitive family. Variables associated with assay conditions will dictate the proper working dilution.
Wange, R.L., et al. 1991. J. Biol. Chem. 266, 11409.
Pang, I-H., and Sternweis, P.C. 1990. J. Biol. Chem. 265, 18707.
Strathmann, M., et al. 1989. Proc. Natl. Acad. Sci. USA86, 7407.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen Hu et al.
The Journal of biological chemistry, 287(17), 13911-13918 (2012-03-06)
Appropriate termination of the phototransduction cascade is critical for photoreceptors to achieve high temporal resolution and to prevent excessive Ca(2+)-induced cell toxicity. Using a genetic screen to identify defective photoresponse mutants in Drosophila, we isolated and identified a novel Gα(q)
Timo Littmann et al.
Analytical biochemistry, 573, 8-16 (2019-03-12)
Functional selectivity of agonists has gained increasing interest in G protein-coupled receptor (GPCR) research, e.g. due to expectations of drugs with reduced adverse effects. Different agonist-dependent GPCR conformations are conceived to selectively activate a balanced or imbalanced intracellular signalling response
Christine Houston et al.
Journal of neurochemistry, 80(4), 678-696 (2002-02-14)
The human histamine H2-receptor (hH2R) couples to Gs-proteins to activate adenylyl cyclase and to Gq-proteins to activate phospholipase C, but phospholipase C activation has not consistently been observed. The aim of this study was to compare coupling of hH2R to
Joseph Simon et al.
The Journal of biological chemistry, 277(35), 31390-31400 (2002-06-25)
Rat brain capillary endothelial (B10) cells express an unidentified nucleotide receptor linked to adenylyl cyclase inhibition. We show that this receptor in B10 cells is identical in sequence to the P2Y(12) ADP receptor ("P2Y(T)") of platelets. When expressed heterologously, 2-methylthio-ADP
Apurva A Javkhedkar et al.
Physiological reports, 3(11) (2015-11-26)
Previously we have shown that in spontaneously hypertensive rats (SHR) renal angiotensin (Ang) II receptor (AT1R) upregulation leads to overstimulation of Na/K-ATPase by Ang II. There are reports that antioxidants can reduce oxidative stress and blood pressure (BP) in SHR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service