Skip to Content
Merck
All Photos(1)

Key Documents

324761

Sigma-Aldrich

Elastase Inhibitor V

Synonym(s):

Elastase Inhibitor V, 2-(2-Bromophenyl)-5-chloro-3,1-benzoxazin-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H7BrClNO2
Molecular Weight:
336.57
UNSPSC Code:
12352200
NACRES:
NA.54

Assay

≥99% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

off-white

solubility

DMSO: 50 mg/mL (Use only fresh DMSO.)

shipped in

ambient

storage temp.

2-8°C

General description

A benzoxazinone compound that acts as a potent inhibitor of human leukocyte elastase (IC50 = 29.5 nM), likely by covalently modifying active site serine via a Michael addition-elimination reaction. Shown to inhibit medium elastase activity (IC50 = 80.8 nM), but not O2•- generation, in fLMP (Cat. No. 05-22-2500)-stimulated primary human neutrophil cultures. Reported to reduce neutrophil infiltration in lung after hemorrhagic shock in rats in vivo (1 mg/kg, i.v.) as assessed by increase in lung wet/dry ratio (0.75- vs. 1.23-fold increase, respectively, with or without treatment) and MPO activity (1.08- vs. 3.72-fold increase, respectively, with or without treatment).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Elastase
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Hsieh, P.W., et al. 2010. Eur. J. Med. Chem.45, 3111.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service