Skip to Content
Merck
All Photos(1)

Key Documents

324713

Sigma-Aldrich

Endoproteinase Glu-C, Excision Grade, Staphylococcus aureus

Synonym(s):

Endoproteinase Glu-C, Excision Grade, Staphylococcus aureus, V8 Protease

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54

biological source

Staphylococcus aureus

Quality Level

grade

Proteomics Grade

form

lyophilized

specific activity

≥10 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

2-8°C

General description

Native endoproteinase Glu-C from Staphylococcus aureus. Serine protease that specifically hydrolyzes peptide bonds at the carboxylic side of glutamic acid and aspartic acid residues. Inhibited by DFP and α2-macroglobulin. Suggested working concentration: 1:20 to 1:100 (protease:protein by weight) for sequence analysis.

Warning

Toxicity: Harmful (C)

Unit Definition

One unit is defined as the amount of enzyme that will hydrolyze 1.0 µmol Z-Phe-Leu-Glu-pNA per min at 25°C, pH 7.8.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) for long term storage or refrigerate (4°C) for short term storage. Stock solutions are stable for up to 2 days at 4°C or for up to 1 month at -20°C.

Other Notes

Houmard, J., and Drapeau, G.R. 1972. Proc. Natl. Acad. Sci. USA69, 3506.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Zhang et al.
The Journal of biological chemistry, 271(52), 33575-33579 (1996-12-27)
While multiple G protein beta and gamma subunit isoforms have been identified, the implications of this potential diversity of betagamma heterodimers for signaling through betagamma-regulated effector pathways remains unclear. Furthermore the molecular mechanism(s) by which the betagamma complex modulates diverse
Victoria L Stefanelli et al.
Matrix biology : journal of the International Society for Matrix Biology, 82, 86-104 (2019-04-21)
The extracellular matrix (ECM) microenvironment is increasingly implicated in the instruction of pathologically relevant cell behaviors, from aberrant transdifferentation to invasion and beyond. Indeed, pathologic ECMs possess a panoply of alterations that provide deleterious instructions to resident cells. Here we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service