Skip to Content
Merck
All Photos(1)

Key Documents

250255-M

Millipore

Cytochalasin D

from Zygosporium mansonii, ≥98% (TLC), solid, fungal toxin, Calbiochem®

Synonym(s):

Cytochalasin D, Zygosporium mansonii

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H37NO6
CAS Number:
Molecular Weight:
507.62
MDL number:
UNSPSC Code:
12352200

Product Name

Cytochalasin D, Zygosporium mansonii, Cytochalasin D, Zygosporium mansonii, CAS 22144-77-0, is a fungal toxin that is about 10-fold more potent than cytochalasin B in inhibiting actin filament function. Blocks cytoplasmic division.

Quality Level

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 2 mg/mL
chloroform: 2 mg/mL

storage temp.

−20°C

SMILES string

N1[C@H]([C@H]3[C@]4([C@H]([C@@H](C(=C)[C@H]3C)O)\C=C\C[C@@H](C(=O)[C@@](\C=C\[C@H]4OC(=O)C)(O)C)C)C1=O)Cc2ccccc2

InChI

1S/C30H37NO6/c1-17-10-9-13-22-26(33)19(3)18(2)25-23(16-21-11-7-6-8-12-21)31-28(35)30(22,25)24(37-20(4)32)14-15-29(5,36)27(17)34/h6-9,11-15,17-18,22-26,33,36H,3,10,16H2,1-2,4-5H3,(H,31,35)/b13-9+,15-14+/t17-,18+,22-,23-,24+,25-,26+,29+,30+/m0/s1

InChI key

SDZRWUKZFQQKKV-JHADDHBZSA-N

General description

A cell-permeable fungal toxin that is approximately 10 times more potent than cytochalasin B in inhibiting actin filament function. Does not inhibit monosaccharide transport across cell membranes. Inhibits cytoplasmic division by blocking the formation of contractile microfilaments at 10 µM. Inactivates low conductance K+ channels. Also modulates CD4 cross-linking in T lymphocytes and increases intracellular Ca2+ levels. Cytochalasin D also exhibits antibiotic and anti-tumor activity.
Cell-permeable fungal toxin. About 10-fold more potent than cytochalasin B in inhibiting actin filament function. Does not inhibit monosaccharide transport across cell membranes. Inhibits cytoplasmic division by blocking the formation of contractile microfilaments. Inactivates low conductance K+ channels. Also modulates CD4 cross-linking in T lymphocytes and increases intracellular Ca2+ levels. Exhibits antitumor activity. Also available as a 10 mM solution in DMSO (Cat. No. 504776).

Biochem/physiol Actions

Primary Target
Actin filament function

Warning

Toxicity: Highly Toxic & Carcinogenic / Teratogenic (I)

Reconstitution

Following reconstitution in chloroform, aliquot, dry under a stream of nitrogen, and freeze (-20°C). Reconstitute in DMSO just prior to use. Dried aliquots are stable for up to 3 months at -20°C.

Other Notes

Sasaki, H., et al. 1995. Proc. Natl. Acad. Sci. USA 92, 2026.
Blankson, H., et al. 1995. Exp. Cell Res.218, 522.
Melamed, I., et al. 1995. Biochem. Biophys. Res. Commun.209, 1102.
Verschueren, H., et al. 1995. Eur. J. Cell Biol.66, 335.
Aszalos, A., et al. 1994. Cell Immunol. 157, 81.
Wang, W.H., et al. 1994. Am. J. Physiol. 267, F592.
Betina, V., and Micekova, D. 1972. Chem. Abstr.77, 160508q.
Katagiri, K., and Matsuura, S. 1971. J. Antibiot. 24, 722.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service