The protein concentration is determined by measuring the absorbance at 280 nm.
233279
Coagulation Factor IX, Human Plasma
Coagulation Factor IX, Human Plasma, CAS 9001-28-9, is a native coagulation factor IX from human plasma. Sold on the basis of plasma equivalent units. 100 plasma equivalent units = 500 µg.
Synonym(s):
Coagulation Factor IX, Human Plasma, Antihemophilic Factor B, Christmas Factor, Plasma Thromboplastin Component, PTC
About This Item
Recommended Products
biological source
human plasma
Quality Level
description
Merck USA index - 14, 3922
Assay
≥95% (SDS-PAGE)
form
liquid
specific activity
≥150 units/mg protein (clotting assay)
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
shipped in
wet ice
storage temp.
−70°C
General description
Packaging
Warning
Physical form
Preparation Note
Reconstitution
Analysis Note
Legal Information
Disclaimer
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
How is the protein concentration of Product No. 233279, Coagulation Factor IX, determined?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service