Skip to Content
Merck
All Photos(1)

Key Documents

205531

Sigma-Aldrich

CA-074 Me

≥98% (HPLC), solid, Cathepsin B inhibitor, Calbiochem®

Synonym(s):

CA-074 Me, Cathepsin B Inhibitor IV, [L-3- trans-(Propylcarbamoyl)oxirane-2-carbonyl]-L-isoleucyl-L-proline Methyl Ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H31N3O6
Molecular Weight:
397.47
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

CA-074 Me, CA-074 Me, CAS 147859-80-1, is a cell-permeable analog of CA-074 that acts as an irreversible inhibitor of intracellular cathepsin B.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable analog of CA-074 (Cat. No. 205530) that acts as an irreversible inhibitor of intracellular cathepsin B. Reported to inhibit bone resorption in rodent models and shown to inhibit B16 melanoma cell invasion in vitro.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
cathepsin B
Product does not compete with ATP.
Reversible: no
Target IC50: 2.24 nM against cathepsin B

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Sever, N., et al. 2002. Biol. Chem.383, 839.
Authier, F., et al. 1999. J. Biol. Chem.274, 33723.
Hill, P.A., et al. 1994. J. Cell Biochem. 56, 118.
Buttle, D.J., et al. 1992. Arch. Biochem. Biophys. 299, 377.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carolina Duarte et al.
Journal of cellular and molecular medicine, 26(10), 2841-2851 (2022-04-17)
Emerging studies indicate that intracellular eukaryotic ceramide species directly activate cathepsin B (CatB), a lysosomal-cysteine-protease, in the cytoplasm of osteoclast precursors (OCPs) leading to elevated RANKL-mediated osteoclastogenesis and inflammatory osteolysis. However, the possible impact of CatB on osteoclastogenesis elevated by
Michael C Yoon et al.
Biochemistry, 61(4), 228-238 (2022-02-05)
CA-074 is a selective inhibitor of cathepsin B, a lysosomal cysteine protease. CA-074 has been utilized in numerous studies to demonstrate the role of this protease in cellular and physiological functions. Cathepsin B in numerous human disease mechanisms involves its
Qi Feng Lin et al.
Journal of virology, 97(10), e0082823-e0082823 (2023-09-25)
Reoviruses infect many mammals and are widely studied as a model system for enteric viruses. However, most of our reovirus knowledge comes from laboratory strains maintained on immortalized L929 cells. Herein, we asked whether naturally circulating reoviruses possess the same
Katia Fettucciari et al.
Cellular and molecular life sciences : CMLS, 79(8), 442-442 (2022-07-22)
Clostridioides difficile infection (CDI) causes nosocomial/antibiotic-associated gastrointestinal diseases with dramatically increasing global incidence and mortality rates. The main C. difficile virulence factors, toxins A and B (TcdA/TcdB), cause cytopathic/cytotoxic effects and inflammation. We demonstrated that TcdB induces caspase-dependent, mitochondria-independent enteric
Pin Li et al.
EMBO molecular medicine, 14(1), e14511-e14511 (2021-11-16)
In the course of our studies aiming to discover vascular bed-specific endothelial cell (EC) mitogens, we identified leukemia inhibitory factor (LIF) as a mitogen for bovine choroidal EC (BCE), although LIF has been mainly characterized as an EC growth inhibitor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service