Skip to Content
Merck
All Photos(1)

Key Documents

204879

Sigma-Aldrich

Complement C1s, Human, Activated, Two-Chain Form

Complement C1s, Human, Activated, Two-Chain Form, CAS 80295-35-8, is a native, activated form. Activated C1s continues activation via classical pathway by cleaving, and thus activating, C2 and C4.

Synonym(s):

Complement C1s, Human, Activated, Two-Chain Form

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.25

Quality Level

Assay

≥95% (SDS-PAGE)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Native, human, activated C1s complement component. Present in human serum at 31 µg/ml. Non-activated C1s is found in circulating blood plasma as a Ca2+-dependent dimer in association with one C1q molecule and two C1r molecules to form the first component of complement (C1). Ensuing C1q binding to classical pathway activators, each C1r protein is cleaved to form activated C1r enzyme. Dimeric C1r enzyme cleaves, and thus activates, each C1s molecule into two disulfide-linked fragments of M.W. 59,000 and 28,000. The 28,000 peptide contains the C1s enzymatic active site. Activated C1s continues activation via the classical pathway by cleaving, and thus activating, C2 and C4.
Native, human, activated C1s complement component. Present in human serum at 31 µg/ml. Nonactivated C1s is found in circulating blood plasma as a Ca2+-dependent dimer in association with one C1q molecule and two C1r molecules to form the first component of complement (C1). Ensuing C1q binding to classical complement pathway activators, each C1r protein is cleaved to form activated C1r enzyme. Dimeric C1r enzyme cleaves, and thus activates, each C1s molecule into two disulfide-linked fragments of M.W. 59 kDa and 28 kDa. The 28 kDa peptide contains the C1s enzymatic active site. Activated C1s continues activation via classical pathway by cleaving, and thus activating, C2 and C4.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 130 mM NaCl, 50 mM sodium phosphate, pH 7.2.

Preparation Note

Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Cooper, N.R. 1985. Adv. Immunol. 37, 151.
Sim, R.B. 1981. Methods Enzymol. 80, 6.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service