Skip to Content
Merck
All Photos(1)

Key Documents

17-300

Sigma-Aldrich

Ral Activation Assay Kit

Non-radioactive Ral Activation Assay Kit can be used to precipitate Ral-A-GTP from cell lysates & detection by a Ral-A specific antibody.

Synonym(s):

Activation Assay Kit, Ral Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable
affinity binding assay: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... RALA(5898)

Packaging

Kit capacity: 30 assays

Components

Ral Activation Assay Buffer, 5X (Cat.# 20-196)

100X GTPγS, 10mM (Cat.# 20-176)

100X GDP, 100mM (Cat.# 20-177)

Anti-Ral-A

Ral Assay Reagent (Ral BP1, agarose) (Cat.# 14-415)

Quality

Routinely evaluated by affinity precipitation/immunoblot analysis. Ral-A-GTP was precipitated from EGF-stimulated Cos-7 cell lysates. The precipitated Ral-A-GTP was detected by Western immunoblot analysis using anti-Ral A.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2'-Methylseleno-modified oligoribonucleotides for X-ray crystallography synthesized by the ACE RNA solid-phase approach.
Barbara Puffer,Holger Moroder,Michaela Aigner,Ronald Micura
Nucleic Acids Research null
Elena Tchevkina et al.
Oncogene, 24(3), 329-335 (2004-10-07)
RAS oncogenes play a critical role in oncogenic transformation and metastases formation. Here we show that Ha-ras greatly stimulates spontaneous metastatic activity of transformed cells through the Ras/RalGDS/RalA intracellular signaling pathway. Introduction of RalA alone leads to a drastic increase
M C Araujo et al.
Biochemistry, 39(29), 8519-8525 (2000-07-29)
Quenched fluorescence peptides were used to investigate the substrate specificity requirements for recombinant wild-type angiotensin I-converting enzyme (ACE) and two full-length mutants bearing a single functional active site (N- or C-domain). We assayed two series of bradykinin-related peptides flanked by
Nicolas Personnic et al.
The FEBS journal, 281(13), 2977-2989 (2014-05-13)
Ral proteins are small GTPases that play critical roles in normal physiology and in oncogenesis. There is little information on the GTPase-activating proteins (GAPs) that downregulate their activity. Here, we provide evidence that the noncatalytic β subunit of RalGAPα1/2 β
M Foschi et al.
The EMBO journal, 16(21), 6439-6451 (1997-11-14)
Endothelin-1 (ET-1) induces cell proliferation and differentiation through multiple G-protein-linked signaling systems, including p21ras activation. Whereas p21ras activation and desensitization by receptor tyrosine kinases have been extensively investigated, the kinetics of p21ras activation induced by engagement of G-protein-coupled receptors remains

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service