Skip to Content
Merck
All Photos(1)

Key Documents

164599

Sigma-Aldrich

Aminopeptidase N, Rat

Synonym(s):

APN/CD13, aminopeptidase N

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352202
NACRES:
NA.54

Assay

≥90% (SDS-PAGE)

Quality Level

form

liquid

specific activity

≥100 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

dry ice

storage temp.

−70°C

Related Categories

General description

Native aminopeptidase N purified from rat renal brush-border membranes and cleaved with papain to produce soluble enzyme. A single-chain, integral membrane protease that functions to remove N-terminal amino acids from peptides with broad specificity. Aminopeptidase N is believed to play a role in angiogenesis and elevated levels of aminopeptidase N have been reported in patients with various types of cancer and have been correlated with tumor load. Also involved in the final stages of digestion of small peptides in the intestinal brush border.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will hydrolyze 1.0 µmol L-Leu-pNA per min at 37°C, pH 7.2

Physical form

In 200 mM NaCl, 50 mM Tris-HCl, pH 8.0.

Other Notes

Bhagwat, S.V., et al. 2003. Blood101, 1818.
van Hensbergen, Y., et al. 2002. Clin. Cancer Res.8, 3747.
Ishii, K., et al. 2001. Biol. Pharm. Bull.24, 226.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service