Skip to Content
Merck
All Photos(3)

Key Documents

1.46396

Millipore

Fluid D - Ready-to-use Rinse Fluid

bottle capacity 1000 mL, bottle filling volume 1000 mL, closure type, White screw cap with septum and protector, pack of 6 bottles

Synonym(s):

Fluid D, Rinsing Solution, Washing fluid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106214

Agency

EP 2.6.1
JP 4.06
USP 71
71

Quality Level

sterility

sterile; autoclaved

form

liquid

feature

closure type White screw cap with septum and protector
ready-to-use

packaging

pack of 6 bottles

technique(s)

sterile filtration: suitable

bottle capacity

1000 mL

bottle filling volume

1000 mL

capacity

1000 mL

pH

7.3±0.2

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

storage temp.

2-25°C

suitability

nonselective for

General description

Fluid D is a washingsolution for sterility testing of pharmaceutical samples containing lecithin oroil using the membrane filtration method. Rinse the membrane with Fluid D after filtration and provide a suitable mediumto observe microbial growth.

Application

Fluid D is suitablefor rinsing the solutions containing oil or lecithin during sterility testingby membrane filtration. It is also suitable for the removal of antimicrobialactivity of non-sterile products by membrane filtration. The fluid is alsoexcellent for rinsing sterile pathways of devices and is used for rinse methodtesting of medical devices.

Features and Benefits

  • Our ready-to-use media provide the highestlevel of quality and testing confidence. They are formulated and tested to meetthe pharmacopeia requirements.
  • Sterility testing media and rinse solutions aremanufactured in an ISO 9001, environmentally controlled production center. Eachlot undergoes a stringent quality control (QC) procedure, including pH,sterility, and growth promotion tests.
  • Our manufacturing approach ensures the highest level of clarity forour media and rinsing fluids, thus improving accuracy, and reducing the risk ofincorrect interpretation and false results.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service