1.30100
HY-LiTE® 2 Hygiene Monitoring System
For ATP measurement to assess biological contamination, Suitable for quality monitoring of cleaning processes on surfaces and in process fluids during food and beverages production as well as in other Industrial production applications.
Synonym(s):
Luminometer, ATP meter, ATP monitor, ATP system
About This Item
Recommended Products
Agency
complies with DIN 10124
Quality Level
product line
HY-LiTE®
dilution
(Hygiene monitoring)
application(s)
environmental
food and beverages
industrial hygiene
industrial qc
jet fuel
surface monitoring
water monitoring
storage temp.
no temp limit
General description
Each kit contains the following items, all packed inside a padded nylon carrying bag:
- HY-LiTE® 2 luminometer
- Battery holder (holds 4 sealed 1.5V alkaline batteries, size Mignon LR6 AA)
- AC mains power supply with country adapters for UK, EU, NA, and AU
- USB connection cable
- USB stick containing: TREND 2 data management software, User Manual, Safety Data Sheet, and Step-by-Step Guide.
Application
Features and Benefits
- Reliable and robust product
- Easy 5-buttons operation
- Half-automatic lid
- Temperature compensation
- Unique self-check of calibration
- Advanced data management using TREND 2 software
- Runs from AC mains or from standard AA batteries
Linkage
SKU 1.30100.0301 replaced by SKU 1.30100.0001
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Related Content
Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.
Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.
Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.
Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service