Skip to Content
Merck
All Photos(1)

Key Documents

1.19154

Perforated Lid

edge protected with handle for MAS-100® Iso line air sampler, stainless steel lid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NA.74

material

stainless steel lid

Agency

non-compendial

description

with handle

feature

no. of holes 300 autoclavable

manufacturer/tradename

MAS-100 Iso MH®

application(s)

air monitoring
pharmaceutical

compatibility

for use with 90mm Petri Dishes
for use with MAS-100® Iso

storage temp.

no temp limit

General description

The Perforated lid stainless steel, 300 x 0.6 mm holes with edge protection and handle can be used with the MAS-100 ISO NT® and MAS-100 ISO MH® microbial air samplers. For safe and easy handling in isolators with gloves it is provided with a handle. The sieve of the perforated lid is at the core of the viable air sampler. Each hole must have an identical diameter and length to accelerate the aspirated air correctly. The 3mm thickness of the perforated sieve guarantee a well-defined impaction jet. The impaction speed defines the physical sampling efficiency. The cut-off value d50 (the particle size at which 50% of the particles impact in the agar due to inertia and 50% are exhausted with the airstream) must be identical between holes and between lids. That is why intense quality control of the sieves is performed. The sieve pattern with 300 holes of a diameter of 0.6 mm provides an impaction speed of approximately 20 m/s at a flow rate of 100 l/min. This impaction speed allows a high physical efficiency, meaning a sampling efficiency of more the 50% of small particles at a size of less than 1.1 μm (d50, calculated, below 0.8 μm in validation studies) while maintaining the viability of microorganisms during impaction on the agar surface (biological efficiency). The validation of physical and biological efficiency has been independently performed for each individual MAS Air sampler model. The perforated lid can be autoclaved and disinfected with all disinfectants compatible stainless steel in order to avoid false positive results in high grade cleanrooms. The sampling efficiency for an instrument must remain identical, irrespective of the lid used. Our perforated lids allow the exchange of several lids with identical sieve pattern on the MAS air sampler without the need of re-calibration.

Application

The perforated lid with 300 x 0.6 mm holes sieve, edge protection and handle is used for microbial monitoring of ambient air with MAS-100 Iso NT® or MAS-100 Iso MH® microbial air samplers in isolators or RABS. Microbial air sampling devices are extensively used in Pharmaceutical.

Legal Information

MAS-100 is a registered trademark of MBV AG
MAS-100 ISO MH is a registered trademark of MBV AG
MAS-100 ISO NT is a registered trademark of MBV AG

accessory

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service