Skip to Content
Merck
All Photos(2)

Key Documents

1.13358

Supelco

Acetonitrile

≥99.8% (GC), for preparative HPLC, for preparative liquid chromatography, for chromatography, Prepsolv

Synonym(s):

ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
741857
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-835-2
NACRES:
NB.21
grade:
for chromatography
for preparative liquid chromatography
technique(s):
HPLC: suitable
application(s):
food and beverages
pharmaceutical
bp:
81-82 °C (lit.)
vapor pressure:
72.8 mmHg ( 20 °C)
97 hPa ( 20 °C)

Product Name

Acetonitrile, for preparative chromatography Prepsolv

grade

for chromatography
for preparative liquid chromatography

Quality Level

vapor density

1.41 (vs air)

vapor pressure

72.8 mmHg ( 20 °C)
97 hPa ( 20 °C)

description

for preparative HPLC

product line

Prepsolv

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

524 °C
973 °F

expl. lim.

16 %

technique(s)

HPLC: suitable

impurities

≤0.0002 meq/g Alkalinity
≤0.0005 meq/g Acidity
≤0.05% Water

evapn. residue

≤1.0 mg/L

transmittance

220 nm, ≥90%
240 nm, ≥98%

refractive index

n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−45 °C (lit.)

transition temp

flash point 2 °C

density

0.786 g/mL at 25 °C (lit.)

application(s)

food and beverages
pharmaceutical

format

neat

storage temp.

2-30°C

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Prepsolv solvents are tailored to the requirements of preparative HPLC to facilitate scale-up from analytical to preparative separations. With their extremely low evaporation residue (<1 mg/l) and low water content, they ensure optimal protection for columns.

Application


  • Pharmacokinetic and oral bioavailability study of schaftoside in rats.: This research highlights the use of acetonitrile in chromatographic methods for the pharmacokinetic and bioavailability analysis of schaftoside, providing key insights into its metabolic profile, essential for drug development and clinical studies (Lin et al., 2024).

  • Quantification of mangiferin in streptozotocin-induced diabetic rat plasma using UPLC-MS/MS: Pharmacokinetic assessment of Gentiana rhodantha extract after oral administration.: Demonstrating the utility of acetonitrile in liquid chromatography-mass spectrometry, this study offers a robust method for quantifying natural compounds in biological matrices, crucial for therapeutic evaluations (Zhao and Wu Li, 2024).

  • Lewis base adducts of NpCl(4).: Exploring the coordination chemistry of neptunium(IV) chloride with Lewis bases in acetonitrile, this study advances our understanding of actinide chemistry and its potential applications in nuclear waste management and reprocessing technologies (Lopez et al., 2024).

  • Coordinating nature of M(6)L(12) double-stranded macrocycles: co-ligand competition of perchlorate, water, and acetonitrile depending on metal(II) ions.: Investigating the competitive binding of co-ligands in acetonitrile solutions, this research provides valuable insights into the supramolecular chemistry of macrocyclic compounds, pivotal for the design of molecular sensors and catalysts (An et al., 2024).

  • Pharmacokinetic Properties of Baitouweng Decoction in Bama Miniature Pigs: Implications for Clinical Application in Humans.: Utilizing acetonitrile in the preparation and analysis of traditional Chinese medicine, this study assesses the pharmacokinetics of Baitouweng Decoction, aiding in the translation of its medicinal use from traditional applications to modern clinical practice (Xu et al., 2024).

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Purity (GC): ≥ 99.8 %
Identity (IR): conforms
Evaporation residue: ≤ 1.0 mg/l
Water: ≤ 0.05 %
Acidity: ≤ 0.0005 meq/g
Alkalinity: ≤ 0.0002 meq/g
Transmission (at 220 nm): ≥ 90 %
Transmission (from 240 nm): ≥ 98 %
Filtered by 0.2 µm filter

Legal Information

PREPSOLV is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service