Skip to Content
Merck
All Photos(1)

Key Documents

1.09969

Supelco

Lead standard

1000 mg Pb, (Pb(NO₃)₂ in H₂O) Titrisol®

Synonym(s):

Lead standard for aqueous matrices

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

Titrisol®

form

concentrate

concentration

1000 mg Pb in H2O

technique(s)

AAS: suitable
ICP: suitable

pH

3.8 (20 °C in H2O)

density

1.03 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

single component solution

storage temp.

15-25°C

Features and Benefits

  • TitrisolTitrisol® offers a wide range of concentrated single-element standards.
  • Dilute the concentrated solution to achieve the desired standard concentration.
  • Directly traceable to standard reference materials (SRM) from the National Institute of Standards and Technology (NIST).
  • Supplied in easy-to-handle plastic ampoules.
  • Accompanied with a batch-specific certificate of analysis (COA) containing information on the NIST batch and minimum shelf life.
  • Provides reliable and reproducible results.
  • Analysis Note

    content (Pb): 1000 mg / Ampoule
    Determination method: Complexometric titration.
    (traceable to NIST - SRM 682)
    Accuracy of the method: +/- 2 mg

    Other Notes

    Blocked in the US and Canada due to regulatory reasons

    Legal Information

    Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

    Signal Word

    Danger

    Hazard Classifications

    Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Repr. 1A - Skin Sens. 1 - STOT RE 2

    Storage Class Code

    6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

    WGK

    WGK 3

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Customers Also Viewed

    Slide 1 of 1

    1 of 1

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service