Skip to Content
Merck

Skip To

1.08170

Thioacetamide

GR for analysis ACS,Reag. Ph Eur

Synonym(s):

Thioacetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C2H5NS
CAS Number:
Molecular Weight:
75.13
UNSPSC Code:
12352111
EC Index Number:
200-541-4
NACRES:
NA.21
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

ACS reagent

Quality Segment

agency

reag. Ph. Eur.

potency

301 mg/kg LD50, oral (Rat)

pH

5.2 (20 °C, 100 g/L in H2O)

mp

111-114 °C

solubility

163 g/L

density

1.37 g/cm3

bulk density

600 kg/m3

storage temp.

2-30°C

SMILES string

S=C(N)C

InChI

1S/C2H5NS/c1-2(3)4/h1H3,(H2,3,4)

InChI key

YUKQRDCYNOVPGJ-UHFFFAOYSA-N

Analysis Note

Assay (argentometric): ≥ 99.0 %
Identity (IR-spectrum): passes test
Melting point: 111 - 114 °C
Heavy metals (as Pb): ≤ 0.001 %
Fe (Iron): ≤ 0.0005 %
Sulfated ash: ≤ 0.1 %


Still not finding the right product?

Explore all of our products under Thioacetamide


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters