Skip to Content
Merck
All Photos(4)

Key Documents

1.07213

Millipore

Casein Peptone for Biotechnology and Microbiology

Pancreatically Digested, Granulated, Prime, for Biotechnology and Microbiology, 12.0-14.0% Total N

Synonym(s):

Peptone from casein (Tryptone), Tryptone from casein

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106212
EC Index Number:
293-428-4
NACRES:
NA.74

Product Name

Peptone from casein (Tryptone), pancreatically digested, granulated, suitable for microbiology

biological source

bovine milk

Quality Level

Assay

12.0-14.0% amino-nitrogen basis (purity)

form

medium granules

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 1 kg
pkg of 2.5 kg
pkg of 25 kg

pH

6.7-7.7 (5% in water)

solubility

800 g/L

bulk density

490 kg/m3

application(s)

microbiology

storage temp.

15-25°C

General description

Peptone from casein is widely used in industrial microbiology as a nutrient source for the growth of microorganisms in large-scale fermentation processes. Peptone from casein, also known as tryptone or peptic casein peptone, is a water-soluble protein hydrolysate obtained from the pancreatic digestion of casein and used as a nutrient source in microbial culture media. It provides a wide range of nutrients, including amino acids, vitamins, minerals, and peptides, which support the growth of diverse microorganisms. It is particularly useful for the cultivation of fastidious and heterotrophic microbes that require complex nutrient sources.

Application

Peptone from casein is commonly used as an ingredient in many microbial culture media for the growth and identification of bacteria, yeasts and molds from various pharmaceutical, environmental, food and beverage samples.

Features and Benefits

Our Granulated media are:
  • Safer - Significant reduction of inhalation of toxic and allergenic substances resulting in fewer allergic responses. Less contamination of working tools and environment
  • Accurate - No separation of components and no clumping, even under humid or warm conditions, ensure a uniform product with higher performance accuracy
  • Fast - Rapid and uniform dissolution in water
  • Easier - Easier handling and weighing due to the better flow properties. Media do not stick to the flask or container
  • Reliable - Homogenous distribution of ingredients guarantees high reproducibility even if only small amounts of media are used.

Linkage

Replaces: 1072139027

Analysis Note

Appearance: Lightly brownishyellow granulate.
Total nitrogen (N) (Kjeldahl): 12.0 - 14.0 %
Amino nitrogen (as N): 3.0 - 5.0 %
pH-value (5 %; water): 6.7 - 7.7
Ca (Calcium): ≤ 0.1 %
Sulfated ash (800 °C): ≤ 15.0 %
Loss on drying (105 °C): ≤ 6.0 %
Suitability for microbiology: passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service