Skip to Content
Merck
All Photos(1)

Key Documents

1.01945

Supelco

Bromine

extra pure

Synonym(s):

Bromine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
159.81
MDL number:
UNSPSC Code:
41116105
EC Index Number:
231-778-1
NACRES:
NA.21
form:
liquid

vapor pressure

233 hPa ( 20 °C)

Quality Level

form

liquid

kinematic viscosity

0.314 cSt(20 °C)

bp

58.8 °C/1013 hPa

mp

-7.3 °C

solubility

35 g/L

density

3.12 g/cm3 at 20 °C (liquid)

storage temp.

15-25°C

SMILES string

[Br-]

InChI

1S/BrH/h1H/p-1

InChI key

CPELXLSAUQHCOX-UHFFFAOYSA-M

General description

extra pure

Application

Bromine extra pure. CAS 7726-95-6, molar mass 159.81 g/mol.

Analysis Note

Assay (iodometric)≥ 99 %Identitypasses testIodine (I)≤ 0.002 %Organic bromine compoundspasses testNon volatile matter≤ 0.02 %

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service